Skip to Content
Merck
All Photos(3)

Key Documents

282987

Sigma-Aldrich

Sodium 4-hydroxybenzenesulfonate dihydrate

98%

Synonym(s):

4-Hydroxybenzenesulfonic acid sodium salt, Phenol-4-sulfonic acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4SO3Na·2H2O
CAS Number:
Molecular Weight:
232.19
Beilstein:
3803250
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

functional group

sulfonic acid

SMILES string

[Na+].[H]O[H].[H]O[H].Oc1ccc(cc1)S([O-])(=O)=O

InChI

1S/C6H6O4S.Na.2H2O/c7-5-1-3-6(4-2-5)11(8,9)10;;;/h1-4,7H,(H,8,9,10);;2*1H2/q;+1;;/p-1

InChI key

SXONATFNYJWFNK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 4-hydroxybenzenesulfonate dihydrate has been used in the preparation of:
  • series of layered divalent transition metal benzenesulfonates
  • luminescent ladder-like lanthanide coordination polymers of 4-hydroxybenzenesulfonate
  • new polynorbornene ionomers bearing fluorinated pendant benzenesulfonate groups

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and structures of layered metal sulfonate salts.
Kosnic EJ, et al.
New. J. Chem., 32(5), 790-793 (2008)
Ying Zhang et al.
Inflammation, 43(2), 417-424 (2020-01-18)
PNU-282987, the α7 acetylcholine receptor(α7nAchR) agonist, has been repeatedly reported to play a key role in anti-inflammatory action of multiple disease. However, little is known about its effect on LPS-induced intestinal epithelial barrier dysfunction. This study investigated the protective effects
Megan L Stanchfield et al.
Frontiers in molecular biosciences, 7, 200-200 (2020-09-15)
Previous studies from this lab have determined that dedifferentiation of Müller glia occurs after eye drop application of an α7 nicotinic acetylcholine receptor (nAChR) agonist, PNU-282987, to the adult rodent eye. PNU-282987 acts on α7 nAChRs on retinal pigment epithelial
Synthesis and ionic transport of sulfonated ring-opened polynorbornene based copolymers.
Santiago AA., et al.
Polymer, 52(19), 4208-4220 (2011)
Hongmei Xu et al.
BMC pregnancy and childbirth, 19(1), 188-188 (2019-05-30)
Preeclampsia is associated with chronic inflammation. The cholinergic anti-inflammatory pathway regulates systemic inflammation through activating α7 nicotinic acetylcholine receptors (α7nAChR) expressed in monocytes/macrophages. This study aimed to investigate the role of α7nAChR in peripheral blood monocytes in preeclampsia. Peripheral blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service