Skip to Content
Merck
All Photos(2)

Key Documents

393541

Sigma-Aldrich

Gallium(III) acetylacetonate

99.99% trace metals basis

Synonym(s):

Ga(acac)3, Gallium(III) 2,4-pentanedionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3COCH=C(O-)CH3]3Ga
CAS Number:
Molecular Weight:
367.05
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

solid

reaction suitability

core: gallium
reagent type: catalyst

mp

196-198 °C (dec.) (lit.)

SMILES string

CC(=O)\C=C(\C)O[Ga](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Ga/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

ZVYYAYJIGYODSD-LNTINUHCSA-K

General description

Gallium(III) acetylacetonate is a sol-gel precursor used to synthesize high-purity GaN nano-wires, nano-needles, and gallium oxide thin films, which are relevant to applications in batteries, fuel cells, and solar cells.

Application

Gallium(III) acetylacetonate can be used:
  • As a precursor to synthesize nanocrystalline gallium oxide spinels via solvothermal process for various applications like photocatalysis, battery cathode materials and electrocatalysis.
  • To fabricate LiGa alloy layer on Li metal anode from in-situ electroreduction. It suppresses the anode dendrite formation in lithium-sulfur batteries.
  • To prepare highly efficient gallium-platinum (GaPt3) nanoparticles hot-solvent synthesis which act as electrocatalysts for hydrogen evolution reaction.
  • To synthesize γ-Ga2O3 nanocrystals for fabricating electron-transporting layer for perovskite solar cells. It forms effective interfacial connections with the perovskite top layer, enhancing the efficiency of charge transport.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural Crystallography and Crystal Chemistry
Dymock K and Palenik GJ
Acta Crystallographica Section B, Structural Crystallography and Crystal Chemistry, 30(5), 1364-1366 (1974)
LowTemperature Catalytic Synthesis of Gallium Nitride Nanowires
The Journal of Physical Chemistry B, 106(32), 7796-7799 (2002)
Temperature-controlled catalytic growth of one-dimensional gallium nitride nanostructures using a gallium organometallic precursor
Chang KW,Wu JJ
Applied Physics. A, Materials Science & Processing, 77(6), 769-774 null
Growth of gallium oxide film from gallium acetylacetonate by atomic layer epitaxy.
Nieminen M,et al
Journal of Materials Chemistry, 6, 27-31 null
Matthias Metzger et al.
Analytical and bioanalytical chemistry, 411(19), 4647-4660 (2019-03-09)
The introduction of fluorine into organic molecules leads to new chemical/physical properties. Especially in the field of pharmaceutical as well as technical applications, fluorinated organic substances gain in importance. The OECD identified and categorized 4730 per- and polyfluoroalkyl substances-related CAS

Articles

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service