286842
1-Nonadecanol
99%
Synonym(s):
Nonadecyl alcohol, n-Nonadecanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
99%
mp
60-61 °C (lit.)
functional group
hydroxyl
SMILES string
CCCCCCCCCCCCCCCCCCCO
InChI
1S/C19H40O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20/h20H,2-19H2,1H3
InChI key
XGFDHKJUZCCPKQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
1-Nonadecanol is one of the constituents of supercritical carbon dioxide (SC-CO2) essential oil of freshly collected aerial parts of Heracleum thomsonii (Umbeliferae).
Application
1-Nonadecanol was used as an internal standard in the study of coprostanol in sediments.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Natural product research, 25(13), 1250-1260 (2011-08-23)
Volatile oil composition of hydro-distilled (HD) and supercritical carbon dioxide (SC-CO(2)) essential oil of freshly collected aerial parts of Heracleum thomsonii (Umbeliferae) from the western Himalayas was studied by GC-FID and GC-MS. Results revealed qualitative and quantitative dissimilarity in the
Sedimentary coprostanol in Kaohsiung harbour and the Tan-Shui estuary, Taiwan.
Marine Pollution Bulletin, 28(8), 494-499 (1994)
Food & function, 11(6), 5257-5265 (2020-05-28)
The lipophilic composition of Allium triquetrum L. bulbs, flowers and leaves was studied for the first time by GC-MS. Sixty compounds were firstly identified in A. triquetrum L. Fatty acids represented the major lipophilic family among the studied extracts, with
International journal of molecular sciences, 16(6), 14194-14209 (2015-06-26)
The lipophilic composition of wild Arbutus unedo L. berries, collected from six locations in Penacova (center of Portugal), as well as some general chemical parameters, namely total soluble solids, pH, titratable acidity, total phenolic content and antioxidant activity was studied
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service