Skip to Content
Merck
All Photos(1)

Key Documents

464473

Sigma-Aldrich

2-Hexyl-1-decanol

97%

Synonym(s):

2-Hexyldecanol, 2-Hexyldecyl alcohol, 2-Octyl-1-octanol, Guerbet hexadecanol, Hexyldecanol, Hexyldecyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH[(CH2)5CH3]CH2OH
CAS Number:
Molecular Weight:
242.44
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.449 (lit.)

bp

193-197 °C/33 mmHg (lit.)

mp

−21-−15 °C (lit.)

density

0.836 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCCCC(CO)CCCCCC

InChI

1S/C16H34O/c1-3-5-7-9-10-12-14-16(15-17)13-11-8-6-4-2/h16-17H,3-15H2,1-2H3

InChI key

XULHFMYCBKQGEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hexyl-1-decanol is a branched alcohol. It is one of the constituent of the essential oil, extracted from the roots of Adiantum flabellulatum. It is a long-chain glass forming monohydroxy alcohol and its dielectric relaxation at low temperature has been studied.

Application

2-Hexyl-1-decanol may be employed as an organic solvent to study the extraction of non-polar acidic drugs from human plasma by parallel artificial liquid membrane extraction (PALME).

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

318.2 °F - closed cup

Flash Point(C)

159 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mercedes Roldán-Pijuán et al.
Analytical and bioanalytical chemistry, 407(10), 2811-2819 (2015-02-16)
The new sample preparation concept "Parallel artificial liquid membrane extraction (PALME)" was evaluated for extraction of the acidic drugs ketoprofen, fenoprofen, diclofenac, flurbiprofen, ibuprofen, and gemfibrozil from human plasma samples. Plasma samples (250 μL) were loaded into individual wells in
Deepak Jadhav et al.
ChemSusChem, 10(11), 2527-2533 (2017-04-14)
Growing concern with the effects of CO
Composition of the essential oil of Adiantum flabellulatum.
Kang WY, et al.
Chemistry of Natural Compounds, 45(4), 575-577 (2009)
Yanqin Gao et al.
The Journal of chemical physics, 139(16), 164504-164504 (2013-11-05)
The dielectric relaxation of two long-chain glass forming monohydroxy alcohols, 2-butyl-1-octanol and 2-hexyl-1-decanol, is studied at low temperature. Remarkable broadening from the pure Debye relaxation is identified for the slowest dynamics, differing from the dielectric spectra of short-chain alcohols. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service