Skip to Content
Merck
All Photos(1)

Key Documents

44861

Sigma-Aldrich

1-Eicosanol

technical, ≥90% (GC)

Synonym(s):

Arachinyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)19OH
CAS Number:
Molecular Weight:
298.55
Beilstein:
1705104
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.4 (vs air)

Quality Level

vapor pressure

<0.1 mmHg ( 20 °C)

grade

technical

Assay

≥90% (GC)

mp

62-65 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCCCCCCCCCCCCCCCO

InChI

1S/C20H42O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h21H,2-20H2,1H3

InChI key

BTFJIXJJCSYFAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

381.2 °F - open cup

Flash Point(C)

194 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vera M S Isca et al.
Food chemistry, 165, 330-336 (2014-07-21)
Salicornia ramosissima J. Woods is considered, in the Iberian Peninsula and France, a gourmet product. Nevertheless, is one of the less studied Salicornia species. In this work, GC-MS was employed to, for the first time; fully characterise the lipophilic profile
Carlos Rogério Figueiredo et al.
Advanced pharmaceutical bulletin, 4(Suppl 1), 429-436 (2014-11-05)
The antitumor activity of Kielmeyera coriacea (Clusiaceae), a medicinal plant used in the treatment of parasitic, as well as fungal and bacterial infections by the Brazilian Cerrado population, was investigated. A chloroform extract (CE) of K. coriacea was tested in
Marc Plante et al.
Methods in molecular biology (Clifton, N.J.), 579, 469-482 (2009-09-19)
A gradient HPLC-charged aerosol detection method was applied to the measurement of different lipids including: free fatty acids, fatty alcohols, glycerides, steroids, phospholipids, and fat-soluble vitamins. An algal oil sample is used as an example. Charged aerosol detection offers a
Polymorphism of n-alkanols: 1-Heptadecanol, 1-octadecanol, 1-nonadecanol, and 1-eicosanol.
Ventola L, et al.
Chemistry of Materials, 14(2), 508-517 (2002)
Ernesto J Méndez Antolín et al.
Journal of AOAC International, 91(5), 1013-1019 (2008-11-05)
A new validated gas chromatographic (GC) method using acetyl chloride as a derivatizing agent for determining policosanol's high molecular weight alcohols (policosanol's fatty alcohols; PFAs) in fish oil (FO) is described. Before derivatizing the alcohols, FO was subjected to a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service