241695
1-Heptadecanol
98%
Synonym(s):
Heptadecyl alcohol
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
Quality Level
Assay
98%
mp
51-55 °C (lit.)
solubility
chloroform: soluble 50 mg/mL, clear, colorless
functional group
hydroxyl
SMILES string
CCCCCCCCCCCCCCCCCO
InChI
1S/C17H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18/h18H,2-17H2,1H3
InChI key
GOQYKNQRPGWPLP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The polymorphism of 1-heptadecanol was studied by differential scanning calorimetry (DSC), X-ray powder diffraction, Raman scattering and infrared spectroscopy (IR).
Application
- Antidandruff activity and chemical constituents of the leaves of Betula cylindrostachya Lindl. ex Wall: This study highlights the isolation of 1-heptadecanol among other compounds from the leaves, identifying its potential in antidandruff applications (C Singla, M Ali, 2018).
- Preparation of Sodium Heptadecyl Sulfate (Tergitol-7i): Describes the synthesis process of sodium heptadecyl sulfate from 1-heptadecanol, underlining its importance in industrial applications (BA Banasik, M Samadpour, 2017).
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
309.2 °F - closed cup
Flash Point(C)
154 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Polymorphism of n-alkanols: 1-Heptadecanol, 1-octadecanol, 1-nonadecanol, and 1-eicosanol.
Chemistry of Materials, 14(2), 508-517 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service