Skip to Content
Merck
All Photos(3)

Key Documents

248592

Sigma-Aldrich

Bismuth(III) nitrate pentahydrate

reagent grade, 98%

Synonym(s):

Bismuth trinitrate pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi(NO3)3 · 5H2O
CAS Number:
Molecular Weight:
485.07
EC Number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Quality Level

Assay

98%

form

crystals

reaction suitability

core: bismuth
reagent type: catalyst

bp

75-80 °C (lit.)

mp

30 °C (lit.)

SMILES string

O=[N+](O[Bi](O[N+]([O-])=O)O[N+]([O-])=O)[O-].C

InChI

1S/CH4.Bi.3NO3/c;;3*2-1(3)4/h1H4;;;;/q;+3;3*-1

InChI key

DPLOIMSBRDLLRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bismuth(III) nitrate pentahydrate is the hydrated form of bismuth(III) nitrate. It is effectively used as a Lewis acid catalyst in organic synthesis for the conversion of thiocarbonyls to their carbonyl compounds; aromatic nitration; protection of carbonyl compounds; Michael reactions, and synthesis of coumarins.

Application

An excellent oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. Convenient reagent for selective oxidation of sufides to sulfoxides.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mashraqui, S.H. et al.
Synthetic Communications, 28, 939-939 (1998)
Mashraqui, S.H. Karnik, M.A.
Synthesis, 713-713 (1998)
Na Li et al.
Dalton transactions (Cambridge, England : 2003), 43(36), 13742-13750 (2014-08-12)
The xFe2O3/yBiOCl composites (xFe/yBi, x/y = 0/100, 5/100, 10/100, 20/100, 30/100 and 40/100 molar ratios) are prepared for the first time through an in situ hydrolysis method under hydrothermal conditions. The samples are characterized by X-ray diffraction (XRD), scanning electron
Nahid Gharib Naseri et al.
Analytical and bioanalytical chemistry, 391(4), 1283-1292 (2008-03-21)
Catalytic adsorptive stripping voltammetry (CAdSV) has been demonstrated at a bismuth film electrode (BiFE) in an injection-moulded electrochemical micro-flow cell. The polystyrene three-electrode flow cell was fabricated with electrodes moulded from a conducting grade of polystyrene containing 40% carbon fibre
Debasish Bandyopadhyay et al.
European journal of medicinal chemistry, 89, 851-862 (2014-12-03)
Polycyclic aromatic hydrocarbons are widespread in nature with a toxicity range from non-toxic to extremely toxic. A series of pyrenyl derivatives has been synthesized following a four-step strategy where the pyrene nucleus is attached with a basic heterocyclic moiety through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service