Skip to Content
Merck
All Photos(2)

Key Documents

307610

Sigma-Aldrich

Bismuth(III) oxychloride

98%

Synonym(s):

Bismuth(III) chloride oxide, Bismuthyl(III) chloride, Chlorobismuth oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiClO
CAS Number:
Molecular Weight:
260.43
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

solubility

H2O: insoluble(lit.)

density

7.72 g/mL at 25 °C (lit.)

SMILES string

Cl[Bi]=O

InChI

1S/Bi.ClH.O/h;1H;/q+1;;/p-1

InChI key

GLQBXSIPUULYOG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R L Ward et al.
Antimicrobial agents and chemotherapy, 27(3), 306-308 (1985-03-01)
Bismuth salts inhibited plaque formation of all enteric viruses tested, which included four strains of rotavirus and one strain each of echovirus, reovirus, and poliovirus. The compounds had no direct virucidal effect at concentrations ranging from 0.025 to 2.5 mg/ml
Sihai Cao et al.
Nanotechnology, 20(27), 275702-275702 (2009-06-18)
A novel BiOCl film with flowerlike hierarchical structures has been fabricated for the first time by dipping Bi film in a mixed solution of H(2)O(2) and HCl. This method presents the advantages of a simple technique, template-free, uniform and controllable
D W Gump et al.
Medical microbiology and immunology, 181(3), 131-143 (1992-01-01)
The effects of sublethal concentrations of bismuth salts on bacterial invasion of mammalian cells were investigated. Pepto-Bismol, bismuth subsalicylate, and bismuth oxychloride, produced by interacting bismuth subsalicylate and simulated gastric juice, in suspension at concentrations as low as 1.4 mM
Dong-Hong Wang et al.
Nanoscale, 4(24), 7780-7785 (2012-11-10)
Porous BiOCl micro-flowers constructed from ultrathin nanosheets with nearly 100% {001} facets exposed were selectively prepared. The exposed {001} facets terminated with a high density of oxygen atoms and are not only favorable for the adsorption of the cationic dye
Mohammed A Gondal et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 47(8), 1192-1200 (2012-04-18)
BiOCl-assisted photodegradation of Rhodamine B (Rh. B) molecules was investigated by using white light and green pulsed laser as sources of irradiation in the visible region for the first time. The dependences of removal efficiencies on catalyst dosage, incident pulsed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service