Skip to Content
Merck
All Photos(4)

Key Documents

310646

Sigma-Aldrich

Bismuth(III) subnitrate

98%

Synonym(s):

Bismuth(III) nitrate basic, Bismuth(III) oxynitrate, Bismuthyl nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi5O(OH)9(NO3)4
CAS Number:
Molecular Weight:
1461.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

Quality Level

Assay

98%

form

powder or crystals

SMILES string

O[Bi]=O.O[Bi](O)O[N+]([O-])=O.O[Bi](O)O[N+]([O-])=O.O[Bi](O)O[N+]([O-])=O.O[Bi](O)O[N+]([O-])=O

InChI

1S/5Bi.4NO3.9H2O.O/c;;;;;4*2-1(3)4;;;;;;;;;;/h;;;;;;;;;9*1H2;/q+1;4*+3;4*-1;;;;;;;;;;/p-9

InChI key

OAVDTZZNYFIIGB-UHFFFAOYSA-E

Looking for similar products? Visit Product Comparison Guide

Application

Bismuth(III) subnitrate can be used as a catalyst for the acetalization and ketalization of aldehydes and ketones with diols.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bismuth subnitrate as an efficient heterogeneous catalyst for acetalization and ketalization of carbonyl compounds with diols
Wu S, et al.
Catalysis Letters, 124, 127-132 (2008)
Agnete Larsen et al.
Basic & clinical pharmacology & toxicology, 97(3), 188-196 (2005-09-01)
Bismuth is used for a multitude of industrial purposes and has partly replaced toxic heavy metals such as lead and mercury in e.g. lubricants and shotgun pellets. In medicine, bismuth-compounds have long been used to remedy gastrointestinal disorders; lately in
Meredin Stoltenberg et al.
Journal of applied toxicology : JAT, 22(2), 111-115 (2002-03-29)
The consumption of bismuth is increasing and knowledge of the potential teratogenic and reproductive damage of bismuth exposure is fragmentary. In the present study we used autometallography (AMG)--the histochemical silver amplification technique--to trace bismuth in the testis and pituitary glands
M Stoltenberg et al.
Reproductive toxicology (Elmsford, N.Y.), 14(1), 65-71 (2000-02-26)
The histochemical silver amplification technique autometallography (AMG), was used to trace bismuth in the testis of Wistar rats injected intraperitoneally with bismuth subnitrate. In the seminiferous tubules, bismuth was located in lysosomes of Sertoli cells closely associated with heads of
M Stoltenberg et al.
European journal of histochemistry : EJH, 51(1), 53-57 (2007-06-06)
Bismuth - sulphur quantum dots can be silver enhanced by autometallography (AMG). In the present study, autometallographic silver enhanced bismuth-sulphur nanocrystals were isolated from unfixed cryo-sections of kidneys and livers of rats exposed to bismuth (Bi207) subnitrate. After being subjected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service