Skip to Content
Merck
All Photos(3)

Key Documents

550787

Sigma-Aldrich

Vanadyl acetylacetonate

98%

Synonym(s):

VO(acac)2, Vanadium(IV)-oxy acetylacetonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OV(C5H7O2)2
CAS Number:
Molecular Weight:
265.16
Beilstein:
4138236
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

mp

235 °C (dec.) (lit.)

SMILES string

CC(=O)\C=C(\C)O[V](=O)O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.O.V/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;;/q;;;+2/p-2/b2*4-3-;;

InChI key

JFHJZWAQYMGNBE-SUKNRPLKSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Vanadyl acetylacetonate may be used as a precursor for the preparation of vanadium dioxide thin films for applications in “intelligent” window coating and data storage.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doped and un-doped vanadium dioxide thin films prepared by atmospheric pressure chemical vapour deposition from vanadyl acetylacetonate and tungsten hexachloride: the effects of thickness and crystallographic orientation on thermochromic properties.
Binions R,et al.
Journal of Materials Chemistry, 17, 4652-4660 (2007)
Xinjian Shi et al.
Physical chemistry chemical physics : PCCP, 18(13), 9255-9261 (2016-03-16)
WO3-BiVO4 n-n heterostructures have demonstrated remarkable performance in photoelectrochemical water splitting due to the synergistic effect between the individual components. Although the enhanced functional capabilities of this system have been widely reported, in-depth mechanistic studies explaining the carrier dynamics of
On the effects of electric fields in aerosol assisted chemical vapour deposition reactions of vanadyl acetylacetonate solutions in ethanol
Warwick MEA and Binions R
Journal of Nanoscience and Nanotechnology, 11(9), 8126-8131 (2011)
Jack S Shamie et al.
ChemSusChem, 10(3), 533-540 (2016-11-20)
In this study, a new mechanism for the reduction of vanadyl acetylacetonate, VO(acac)
Szymon Kowalski et al.
International journal of molecular sciences, 20(2) (2019-01-13)
Pancreatic cancer is characterized by one of the lowest five-year survival rates. In search for new treatments, some studies explored several metal complexes as potential anticancer drugs. Therefore, we investigated three newly synthesized oxidovanadium(IV) complexes with 2-methylnitrilotriacetate (bcma3-), N-(2-carbamoylethyl)iminodiacetate (ceida3-)

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service