Skip to Content
Merck
All Photos(1)

Key Documents

728594

Sigma-Aldrich

5-Iodo-1-pentyne

≥98.0% (GC)

Synonym(s):

1-Iodo-4-pentyne, 5-Iodo-1-pentyne, 5-Iodopentyne

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7I
CAS Number:
Molecular Weight:
194.01
Beilstein:
1736711
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

liquid

functional group

alkyl halide
iodo

storage temp.

2-8°C

SMILES string

ICCCC#C

InChI

1S/C5H7I/c1-2-3-4-5-6/h1H,3-5H2

InChI key

SEHFYZRHGUPLSY-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

135.3 °F

Flash Point(C)

57.38 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service