Skip to Content
Merck
All Photos(1)

Key Documents

238279

Sigma-Aldrich

1-Iodohexadecane

contains copper as stabilizer, 95%

Synonym(s):

Hexadecyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15I
CAS Number:
Molecular Weight:
352.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

contains

copper as stabilizer

refractive index

n20/D 1.4806 (lit.)

bp

206-207 °C/10 mmHg (lit.)

mp

21-23 °C (lit.)

density

1.121 g/mL at 25 °C (lit.)

functional group

alkyl halide
iodo

SMILES string

CCCCCCCCCCCCCCCCI

InChI

1S/C16H33I/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h2-16H2,1H3

InChI key

KMWHQYDMBYABKL-UHFFFAOYSA-N

Related Categories

Application

1-Iodohexadecane was used in the synthesis of alkyl immidazolium surfactants, 1-hexadecyl-imidazolium (Imm-C16) and dihexadecyl-imidazolium (Imm-2C16). It was also used in the chemical synthesis of 1-O-hexadecyl dihydroxyacetone-3-phosphate.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

161.6 - 162.5 °F - closed cup

Flash Point(C)

72.0 - 72.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melt-processable syndiotactic polystyrene/montmorillonite nanocomposites.
Wang ZM, et al.
Journal of Polymer Science. Part B, Polymer Physics, 41(24), 3173-3187 (2003)
Benay Akyon et al.
Environmental science. Processes & impacts, 21(2), 279-290 (2018-11-20)
Hydraulic fracturing generates large volumes of produced water, and treatment of produced water may be necessary for disposal or reuse. Biological treatment of produced water is a potential approach to remove organic constituents and reduce fouling, in conjunction with other
Chemical synthesis of 1-O-alkyl and 1-O-acyl dihydroxyacetone-3-phosphate.
Hajra A, et al.
Chemistry and Physics of Lipids, 33(2), 179-193 (1983)
Acute toxicities to larval rainbow trout of representative compounds detected in Great Lakes fish.
C C Edsall
Bulletin of environmental contamination and toxicology, 46(2), 173-178 (1991-02-01)
Vinay Kumar Banka et al.
Nuclear medicine and biology, 42(3), 317-322 (2014-12-30)
A lipiodol solution of (188)Re-4-hexadecyl-2,2,9,9-tetramethyl-4,7-diaza-1,10-decanedithiol (HTDD) has been successfully developed for liver cancer therapy; however, its preparation requires a multi-step synthesis and it is characterized by a low labeling yield. We synthesized a new compound, 4-hexadecyl-4,7-diaza-1,10-decanedithioacetate (AHDD), without gem dimethyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service