Skip to Content
Merck
All Photos(2)

Key Documents

233226

Sigma-Aldrich

2,4-Difluoronitrobenzene

99%

Synonym(s):

2,4-Difluoro-1-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3NO2
CAS Number:
Molecular Weight:
159.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

99%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

203-204 °C (lit.)

mp

9-10 °C (lit.)

density

1.451 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

[O-][N+](=O)c1ccc(F)cc1F

InChI

1S/C6H3F2NO2/c7-4-1-2-6(9(10)11)5(8)3-4/h1-3H

InChI key

RJXOVESYJFXCGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nucleophilic aromatic substitution of 2,4-difluoronitrobenzene with morpholine has been investigated using flow reactor with simulated moving bed (SMB) chromatography module.

Application

2,4-Difluoronitrobenzene has been used in the synthesis of:
  • 2,4-difluoro-5-nitrobenzenesulfonic acid via sulfonation reaction
  • (±)-horsfiline
  • resin-bound 2-arylaminobenzimidazoles

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander G O'Brien et al.
Angewandte Chemie (International ed. in English), 51(28), 7028-7030 (2012-06-08)
Continuous synthesis meets continuous purification to produce pure products from crude reaction mixtures. In the nucleophilic aromatic substitution of 2,4-difluoronitrobenzene with morpholine the desired monosubstituted product can be continuously separated from the byproducts in a purity of over 99 %
U M Kremer
Biological chemistry Hoppe-Seyler, 371(9), 861-864 (1990-09-01)
2,4-Difluoro-5-nitrobenzenesulfonic acid has been synthesized by the sulfonation of 2,4-difluoronitrobenzene, and precipitated with KCl as the potassium sulfonate. The structure was confirmed by chemical and spectroscopic methods (IR, 1H-NMR, 13C-NMR, 19F-NMR, UV, MS and ultimate organic analysis). Lysozyme was cross-linked
A direct synthesis of 2-arylpropenoic acid esters having nitro groups in the aromatic ring: a short synthesis of (?)-coerulescine and (?)-horsfiline.
Selvakumar N, et al.
Tetrahedron Letters, 43(50), 9175-9178 (2002)
A solid phase traceless synthesis of 2-arylaminobenzimidazoles.
Krchnak V, et al.
Tetrahedron Letters, 42(9), 1627-1630 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service