261807
2,4,6-Trifluoronitrobenzene
98%
Synonym(s):
1,3,5-Trifluoro-2-nitrobenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
F3C6H2NO2
CAS Number:
Molecular Weight:
177.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.477 (lit.)
bp
182-185 °C (lit.)
mp
3.5 °C (lit.)
density
1.514 g/mL at 25 °C (lit.)
functional group
fluoro
nitro
SMILES string
[O-][N+](=O)c1c(F)cc(F)cc1F
InChI
1S/C6H2F3NO2/c7-3-1-4(8)6(10(11)12)5(9)2-3/h1-2H
InChI key
PWRFDGYYJWQIAB-UHFFFAOYSA-N
Application
2,4,6-Trifluoronitrobenzene has been used in preparation of 4-methoxy-2,6-difluoroanilines, versatile building blocks in medicinal chemistry.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
172.4 °F - closed cup
Flash Point(C)
78 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A convenient synthesis of 4-alkoxy-and 4-hydroxy-2, 6-difluoroanilines.
Alonso-Alija C, et al.
Tetrahedron Letters, 45(1), 95-98 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service