Skip to Content
Merck
All Photos(1)

Key Documents

335789

Sigma-Aldrich

1-Chloro-2,4-difluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3F2
CAS Number:
Molecular Weight:
148.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.475 (lit.)

bp

127 °C (lit.)

density

1.353 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Fc1ccc(Cl)c(F)c1

InChI

1S/C6H3ClF2/c7-5-2-1-4(8)3-6(5)9/h1-3H

InChI key

AJCSNHQKXUSMMY-UHFFFAOYSA-N

Application

1-Chloro-2,4-difluorobenzene was used in the preparation of series of benzonorbornadienes. It was also used in the preparation of difluoroarenes.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K C Caster et al.
The Journal of organic chemistry, 66(9), 2932-2936 (2001-04-28)
This report details the synthesis of several benzonorbornadienes by Diels--Alder cycloaddition of cyclopentadiene derivatives with substituted benzyne intermediates, which were generated by low-temperature metal--halogen exchange of halobenzenes. General conditions were developed, allowing synthesis of most benzonorbornadienes described herein at the
Jason T Manka et al.
The Journal of organic chemistry, 69(6), 1967-1971 (2004-04-03)
Aryldiazo substituents were used in nucleophilic aromatic substitution reactions of halogens. The Ph-N=N- group activates ortho fluorine atoms toward alkylthiolation under mild conditions. In contrast, the Me(2)N-C(6)H(4)-N=N- group has virtually no activation effect in nucleophilic aromatic substitution, and serves as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service