Skip to Content
Merck
All Photos(1)

Key Documents

196622

Sigma-Aldrich

2,5-Difluoronitrobenzene

97%

Synonym(s):

1,4-Difluoro-2-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3NO2
CAS Number:
Molecular Weight:
159.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.509 (lit.)

bp

206.5 °C (lit.)

mp

−11.7 °C (lit.)

density

1.467 g/mL at 25 °C (lit.)

functional group

fluoro
nitro

SMILES string

[O-][N+](=O)c1cc(F)ccc1F

InChI

1S/C6H3F2NO2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H

InChI key

XNJAYQHWXYJBBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Difluoronitrobenzene was used in the synthesis of :
  • N-alkylated 2-arylaminobenzimidazoles
  • quinoxalinones
  • N-(2-nitro-4-fluorophenyl)-l,2,3,4-tetrahydroisoquinoline

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H E Haenen et al.
Chemico-biological interactions, 98(2), 97-112 (1995-11-17)
Proximal tubular biotransformation of the glutathionyl (GSH) conjugate derived from 2,5-difluoronitrobenzene (5-fluoro-2-glutathionyl-nitrobenzene) was studied by means of 19F-NMR. This method allows a direct and specific detection of the fluorinated metabolites formed, at a detection limit of 1 microM for an
Manuel A V Ribeiro da Silva et al.
The journal of physical chemistry. B, 114(40), 12914-12925 (2010-09-24)
This work reports the experimental and computational thermochemical study performed on three difluorinated nitrobenzene isomers: 2,4-difluoronitrobenzene (2,4-DFNB), 2,5-difluoronitrobenzene (2,5-DFNB), and 3,4-difluoronitrobenzene (3,4-DFNB). The standard (p° = 0.1 MPa) molar enthalpies of formation in the liquid phase of these compounds were
A solid phase traceless synthesis of quinoxalinones.
Krchnak V, et al.
Tetrahedron Letters, 41(16), 2835-2838 (2000)
Ring-opening reactions of N-aryl-, 1, 2, 3, 4-tetrahydroisoquinoline derivatives.
Andrew HK and Stanforth SP.
Tetrahedron, 48(4), 743-750 (1992)
A solid phase traceless synthesis of 2-arylaminobenzimidazoles.
Krchnak V, et al.
Tetrahedron Letters, 42(9), 1627-1630 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service