Skip to Content
Merck
All Photos(2)

Key Documents

216178

Sigma-Aldrich

4,4′-Thiodiphenol

99%

Synonym(s):

4-Hydroxyphenyl sulfide, TDP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S(C6H4OH)2
CAS Number:
Molecular Weight:
218.27
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

solid

mp

154-156 °C (lit.)

SMILES string

Oc1ccc(Sc2ccc(O)cc2)cc1

InChI

1S/C12H10O2S/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8,13-14H

InChI key

VWGKEVWFBOUAND-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Materials for the establishment of maximum allowable concentrations of bis (4-oxyphenyl) sulfide in the working zone air].
L A Antonovich et al.
Gigiena truda i professional'nye zabolevaniia, (1)(1), 31-32 (1991-01-01)
Bingli Lei et al.
Chemosphere, 169, 204-211 (2016-11-24)
Many studies have been shown that environmental estrogen bisphenol A (BPA) can activate nuclear receptor (estrogen receptor alpha, ERα) or membrane receptor (G-protein-coupled receptor, GPR30) in breast cancer cells and exerts genomic or nongenomic actions inducing cell proliferation. 4,4'-thiodiphenol (TDP)
Andrey Galukhin et al.
Molecules (Basel, Switzerland), 24(1) (2019-01-10)
We described a synthetic approach to bisphenol-based monocyanate esters based on mono-O-methylation of parental bisphenols followed by cyanation of the residual phenolic hydroxyl. Structures of the synthesized compounds were determined by the application of IR, NMR ¹H and 13C spectroscopies
Lei Wang et al.
Environmental science & technology, 53(12), 7095-7102 (2019-05-28)
Bisphenol A (BPA) is an important endocrine disrupting chemical. Although high levels of BPA in some new clothes have been reported, the occurrence of bisphenol chemicals including BPA in daily clothes is still unknown, and the human exposure to BPA
Katra Kolšek et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(1), 8-15 (2014-09-07)
A homeostasis of the glucocorticoid and androgen endocrine system is essential to human health. Their disturbance can lead to various diseases, for example cardiovascular, inflammatory and autoimmune diseases, infertility, cancer. Fifteen widely used industrial chemicals that disrupt endocrine activity were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service