Skip to Content
Merck
All Photos(2)

Key Documents

436372

Sigma-Aldrich

4,4′-Diaminodiphenyl sulfide

98%

Synonym(s):

4,4′-Thiodianiline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S(C6H4NH2)2
CAS Number:
Molecular Weight:
216.30
Beilstein:
1875513
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

105-107 °C (lit.)

SMILES string

Nc1ccc(Sc2ccc(N)cc2)cc1

InChI

1S/C12H12N2S/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H,13-14H2

InChI key

ICNFHJVPAJKPHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Diaminodiphenyl sulfide is a diaminodiphenyl analog. On oral administration to rats, it induces haematological and pathological changes indicative of erythrocyte destruction. It has been reported to generate hydrogen peroxide in erythrocytes in vitro. Enzymatic oxidation of 4,4′-diaminodiphenyl sulfide to their sulfoxide derivatives in guinea pig liver homogenates has been reported.

Application

4,4′-Diaminodiphenyl sulfide may be employed for the fabrication of quantum wires and quantum dots by chemical vapor deposition.
It may be used for the preparation of the following:
  • polypyromellitimides
  • sulfur-containing copolyimides
  • polyamides

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Munday et al.
Journal of applied toxicology : JAT, 5(6), 414-417 (1985-12-01)
Diphenyl disulphide, 4,4'-diaminodiphenyl disulphide, 2,2'-diaminodiphenyl disulphide, 4,4'-dimethyldiphenyl disulphide and 4,4'-dinitrodiphenyl disulphide, when administered orally to rats, induced haematological and pathological changes indicative of erythrocyte destruction in vivo. No evidence of haemolysis was detected, however, in animals receiving diphenyl disulphide-2,2'-dicarboxylic acid
The enzymatic formation of sulfoxides: the oxidation of chlorpromazine and 4,4'-diaminodiphenyl sulfide by guinea pig liver microsomes.
J R GILLETTE et al.
The Journal of pharmacology and experimental therapeutics, 130, 262-267 (1960-11-01)
Quantum wire and dot formation by chemical vapor deposition and molecular layer deposition of one-dimensional conjugated polymer.
Yoshimura T, et al.
Applied Physics Letters, 60(3), 268-270 (1992)
R Munday
Journal of applied toxicology : JAT, 5(6), 409-413 (1985-12-01)
Diphenyl disulphide has been shown to generate hydrogen peroxide in erythrocytes in vitro. It also induces oxidative damage (reversible and irreversible haemoglobin oxidation, depletion of non-protein and protein-bound thiols) in these cells. Such changes were also recorded in erythrocytes exposed
Synthesis and characterization of novel, soluble sulfur-containing copolyimides with high refractive indices.
Dusselberg D, et al.
J. Mater. Sci., 46(14), 4872-4879 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service