Skip to Content
Merck
All Photos(2)

Key Documents

168734

Sigma-Aldrich

4,4′-Dihydroxybiphenyl

97%

Synonym(s):

4,4′-Biphenol, 4,4′-Biphenyldiol, 4,4′-Diphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4C6H4OH
CAS Number:
Molecular Weight:
186.21
Beilstein:
1908886
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

powder

mp

280-282 °C (lit.)

SMILES string

Oc1ccc(cc1)-c2ccc(O)cc2

InChI

1S/C12H10O2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8,13-14H

InChI key

VCCBEIPGXKNHFW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Used in the preparation of polyether liquid crystals.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Self-heat. 2 - Skin Sens. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

537.8 °F

Flash Point(C)

281 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angewandte Chemie (International Edition in English), 106, 1563-1563 (1994)
J H Golbeck et al.
Journal of bacteriology, 156(1), 49-57 (1983-10-01)
Biphenyl metabolism in Aspergillus toxicarius occurs by successive hydroxylations in the 4- and 4'-positions, followed by conjugation with sulfate to produce 4-hydroxybiphenyl-O-sulfonic acid and 4,4'-dihydroxybiphenyl-O-sulfonic acid. The hydroxylation reactions normally occur only after a prolonged lag period after which the
Jae Kyung No et al.
Biological & pharmaceutical bulletin, 29(1), 14-16 (2006-01-06)
In our previous study, we showed that 4,4'-dihydroxybiphenyl (44'-BP) reduced melanin content via the inhibition of tyrosinase. In the current study, we utilized 44'-BP treated B16 melanoma cells (B16 cells) to measure several key cellular parameters known to be involved
Takumi Yamaguchi et al.
Chemistry, an Asian journal, 2(4), 468-476 (2007-04-19)
Upon complexation with Pd(II) ions, precisely designed strandlike ligands with two tris(3,5-pyridine) units at both terminals were assembled, with the aid of a linear template molecule, into a discrete tubular complex with a length of 3.5 nm. The high stability
Hydroxylations of biphenyl by fungi.
R V Smith et al.
The Journal of applied bacteriology, 49(1), 65-73 (1980-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service