Skip to Content
Merck
All Photos(3)

Key Documents

428302

Sigma-Aldrich

Polysulfone

average Mw ~35,000 by LS, average Mn ~16,000 by MO, pellets (Transparent)

Synonym(s):

Poly[oxy-1,4-phenylenesulfonyl-1,4-phenyleneoxy-1,4-phenylene(1-methylethylidene)-1,4-phenylene], Polysulfone resin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H4-4-C(CH3)2C6H4-4-OC6H4-4-SO2C6H4-4-O]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets (Transparent)

Quality Level

autoignition temp.

1022 °F

melt index

6.5 g/10 min (343°C/0.3 Mpa)

mol wt

average Mn ~16,000 by MO
average Mw ~35,000 by LS

reduced viscosity

0.37-0.39 dL/g, 0.2 % (w/v) in chloroform(25 °C)(lit.)

density

1.24 g/mL at 25 °C (lit.)

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(O)cc2.Oc3ccc(cc3)S(=O)(=O)c4ccc(O)cc4

Looking for similar products? Visit Product Comparison Guide

Application

Microwave cookware, appliance covers, surgical tools, automotive electrical components, printed circuit boards and plumbing components.

Features and Benefits

High impact resistance. Temperature use range up to 160°C. Exceptional steam sterilization stability. Electrical properties are maintained over a wide temperature and frequency range. High resistance to radiation degradation.

Physical form

Thermoplastic engineering polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M U M Junaidi et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(9), 2102-2109 (2013-05-10)
Although ultrafiltration (UF) membranes are applicable in wastewater and water treatment, most UF membranes are hydrophobic and susceptible to severe fouling by natural organic matter. In this work, polysulfone (PSf) membrane was blended with silicaluminophosphate (SAPO) nanoparticles, SAPO-34, to study
Marinela Z Knezevic et al.
Renal failure, 34(7), 849-855 (2012-05-23)
The quality of life in patients undergoing hemodialysis is significantly disturbed. There are data that hemodiafiltration (HDF) may be more effective than conventional hemodialysis in the removal of uremic toxins and may reduce frequency and severity of intradialytic and postdialysis
Yong Soo Choi et al.
Talanta, 94, 353-355 (2012-05-23)
Ultrapure water produced by a water purification system is one of the most essential and widely used reagents in laboratories. However, its quality is usually the least well-characterized and often overlooked. Here we investigate the contamination of ultrapure water by
John T Daugirdas et al.
Kidney international, 82(2), 147-157 (2012-05-18)
Substantial activation of platelets can occur in the course of hemodialysis. Platelet surface markers show evidence of platelet degranulation. Some activation occurs due to exposure of blood to the roller pump segment and microbubbles may play a role. Platelet activation
Sandra Pérez et al.
The Analyst, 137(16), 3854-3861 (2012-07-06)
A novel amperometric bienzymatic biosensor has been developed based on the incorporation of Lactate Oxidase (LOx) and Horseradish Peroxidase (HRP) into a carbon nanotube/polysulfone membrane by the phase inversion technique onto screen-printed electrodes (SPEs). In order to improve the sensitivity

Articles

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service