Skip to Content
Merck
All Photos(1)

Key Documents

213055

Sigma-Aldrich

4-Chloro-3-nitrotoluene

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)Cl
CAS Number:
Molecular Weight:
171.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

liquid

refractive index

n20/D 1.558 (lit.)

bp

260 °C/745 mmHg (lit.)

mp

7 °C (lit.)

density

1.297 g/mL at 25 °C (lit.)

functional group

chloro
nitro

SMILES string

Cc1ccc(Cl)c(c1)[N+]([O-])=O

InChI

1S/C7H6ClNO2/c1-5-2-3-6(8)7(4-5)9(10)11/h2-4H,1H3

InChI key

NWESJZZPAJGHRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vibrational spectral analysis of 4-chloro-3-nitrotoluene has been studied using Raman and IR spectroscopy.

Application

4-Chloro-3-nitrotoluene was used in the synthesis of 4-(2-hydroxyethylamino)-3-nitrotoluene.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

264.2 °F

Flash Point(C)

129 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel V Labarbera et al.
Bioorganic & medicinal chemistry, 13(2), 387-395 (2004-12-16)
The imidazoquinoxalinones 1 and 2 are benzimidazole analogues of indole-based marine natural products called makaluvamins. The stabilized cation 1 and the zwitterion 2 were prepared in approximately 9 steps from readily available starting materials. Compound 1 is more cytostatic and
M Govindarajan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 89, 137-148 (2012-01-21)
In this work, the vibrational spectral analysis was carried out by using Raman and infrared spectroscopy in the range 100-4000 cm(-1) and 50-4000 cm(-1), respectively, for 4-chloro-3-nitrotoluene (C7H6NO2Cl) molecule. The molecular structure, fundamental vibrational frequencies and intensity of the vibrational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service