Skip to Content
Merck
All Photos(1)

Key Documents

101702

Sigma-Aldrich

4-Chloro-2-nitrotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)Cl
CAS Number:
Molecular Weight:
171.58
Beilstein:
2046651
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

239-240 °C/718 mmHg (lit.)

mp

34-38 °C (lit.)

functional group

chloro

SMILES string

Cc1ccc(Cl)cc1[N+]([O-])=O

InChI

1S/C7H6ClNO2/c1-5-2-3-6(8)4-7(5)9(10)11/h2-4H,1H3

InChI key

SQFLFRQWPBEDHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chloro-2-nitrotoluene can be used in the synthesis of indigo dye.

Biochem/physiol Actions

4-Chloro-2-nitrotoluene is the starting reagent for synthesis of tricyclic indole-2-carboxylic acids, a potential NMDA-glycine antagonists.

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thanasis Karapanayiotis et al.
The Analyst, 129(7), 613-618 (2004-06-24)
The Raman and electron impact mass spectra of synthetic indigo and its four 6,6'-dihalogeno analogues are reported and discussed. The influence of varying the halogen on these Raman spectra is considered. Particular emphasis is laid on distinguishing indigo from 6,6'-dibromoindigo
S Katayama et al.
The Journal of organic chemistry, 66(10), 3474-3483 (2001-05-12)
The practical synthesis of a series of tricyclic indole-2-carboxylic acids, 7-chloro-3-arylaminocarbonylmethyl-1,3,4,5-tetrahydrobenz[cd]indole-2-carboxylic acids, as a new class of potent NMDA-glycine antagonists is described. The synthetic route to the key intermediate 12a comprises a regioselective iodination of 4-chloro-2-nitrotoluene, modified Reissert indole synthesis
D de Almeida Azevedo et al.
Journal of chromatography. A, 879(1), 13-26 (2000-06-28)
Gas chromatography-mass spectrometry (GC-MS) and liquid chromatography-atmospheric pressure chemical ionization mass spectrometry (LC-APCI-MS) were optimized and applied for the trace-level determination of 42 priority pesticides and 33 priority organic pollutants from European Union Directive EC 76/464. First, off-line solid-phase extraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service