Skip to Content
Merck
All Photos(1)

Key Documents

226653

Sigma-Aldrich

5-Chloro-2-nitrotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)Cl
CAS Number:
Molecular Weight:
171.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

27-30 °C (lit.)

functional group

chloro

SMILES string

Cc1cc(Cl)ccc1[N+]([O-])=O

InChI

1S/C7H6ClNO2/c1-5-4-6(8)2-3-7(5)9(10)11/h2-4H,1H3

InChI key

NSMZCUAVEOTJDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The cytotoxicity of 5-chloro-2-nitrotoluene, an analog of chlorphenamidine, was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of chlorphenamidine and its metabolites on HeLa cells.
M Murakami et al.
Bulletin of environmental contamination and toxicology, 11(2), 184-188 (1974-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service