Skip to Content
Merck
All Photos(1)

Key Documents

219428

Sigma-Aldrich

4-Fluoroiodobenzene

99%

Synonym(s):

1-Fluoro-4-iodobenzene, 1-Iodo-4-fluorobenzene, 4-Fluoro-1-iodobenzene, 4-Fluorophenyl iodide, 4-Iodo-1-fluorobenzene, 4-Iodofluorobenzene, p-Fluoroiodobenzene, p-Fluorophenyl iodide, p-Iodofluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4I
CAS Number:
Molecular Weight:
222.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.583 (lit.)

bp

182-184 °C (lit.)

mp

−20 °C (lit.)

density

1.925 g/mL at 25 °C (lit.)

functional group

fluoro
iodo

SMILES string

Fc1ccc(I)cc1

InChI

1S/C6H4FI/c7-5-1-3-6(8)4-2-5/h1-4H

InChI key

KGNQDBQYEBMPFZ-UHFFFAOYSA-N

General description

4-[18F] Fluoroiodobenzene is a versatile building block in 18F radiochemistry and is used in various transition metal-mediated C-C and C-N cross-coupling reactions. Cross-coupling reaction of 4-fluoroiodobenzene with potassium pentafluorophenyltrifluoroborate in the presence of different palladium catalysts and silver oxide in toluene has been investigated.

Application

4-Fluoroiodobenzene was used in the preparation of fluorotriphenylene derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thiruvellore Thatai Jayanth et al.
Chemical communications (Cambridge, England), (8)(8), 894-896 (2006-02-16)
Highly substituted triphenylene derivatives were prepared in good yields via the palladium-catalyzed carbocyclization of arynes with aryl iodides.
The effect of N-heterocyclic carbene ligands in the palladium-catalyzed cross-coupling reaction of K [C6F5BF3] with aryl iodides and aryl bromides.
Adonin NY, et al.
Tetrahedron, 64(25), 5920-5924 (2008)
Jenilee Dawn Way et al.
Journal of labelled compounds & radiopharmaceuticals, 57(2), 104-109 (2014-03-29)
4-[18F]Fluoroiodobenzene ([18F]FIB) is a versatile building block in 18F radiochemistry used in various transition metal-mediated C-C and C-N cross-coupling reactions and [18F]fluoroarylation reactions. Various synthesis routes have been described for the preparation of [18F]FIB. However, to date, no automated synthesis
Gabriel A Odugbesi et al.
Journal of chromatography. A, 1604, 460466-460466 (2019-09-03)
Ionic liquids (ILs) are well-known in the field of separation science for their unique selectivity when used as stationary phases in gas chromatography (GC). While a significant amount of knowledge has been attained in correlating structural features of an IL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service