Skip to Content
Merck
All Photos(3)

Key Documents

178004

Sigma-Aldrich

1-Chloro-2-methylpropane

98%

Synonym(s):

Isobutyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2Cl
CAS Number:
Molecular Weight:
92.57
Beilstein:
635650
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.398 (lit.)

bp

68-69 °C (lit.)

mp

−131 °C (lit.)

solubility

H2O: insoluble
alcohol: miscible
diethyl ether: miscible

density

0.883 g/mL at 25 °C (lit.)

functional group

alkyl halide

storage temp.

2-8°C

SMILES string

CC(C)CCl

InChI

1S/C4H9Cl/c1-4(2)3-5/h4H,3H2,1-2H3

InChI key

QTBFPMKWQKYFLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The gas-solid virial co-effiicent of 1-chloro-2-methylpropane was determined. The infrared and Raman spectra of 1-chloro-2-methylpropane was studied.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.9 °F - closed cup

Flash Point(C)

-19.39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crowder GA, et al.
Journal of Molecular Structure, 64, 193-199 (1980)
Rybolt et al.
Journal of colloid and interface science, 220(1), 148-156 (1999-11-07)
Second gas-solid virial coefficients were determined at 403.5 +/- 0.5 K for 6 adsorbates, including butane, chloroform, trichlorofluoromethane (Freon 11), bromochloromethane, 1-chloro-2-methylpropane, and dibromodifluoromethane. For another 11 adsorbates, including dichlorodifluoromethane (Freon 12), chlorodifluoromethane (Freon 22), methyl chloride, methylene chloride, propane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service