Skip to Content
Merck
All Photos(2)

Key Documents

294861

Sigma-Aldrich

1-Chloroadamantane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15Cl
CAS Number:
Molecular Weight:
170.68
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

165-166 °C (lit.)

functional group

chloro

SMILES string

ClC12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C10H15Cl/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9H,1-6H2/t7-,8+,9-,10-

InChI key

OZNXTQSXSHODFR-CHIWXEEVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An unusual zwitterionic diradical intermediate complex was generated during the photoinduced electron-transfer substitution reaction between 1-chloroadamantane and tert-butylamine system in a hydrocarbon glass.

Application

1-Chloroadamantane was employed as precursor for the synthesis of perfluoroadmantane derivatives via aerosol direct fluorination.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John B Miller et al.
Journal of the American Chemical Society, 126(40), 12742-12743 (2004-10-08)
An unusual zwitterionic diradical intermediate complex R*...X-...HR'B*+ is generated during the photoinduced electron-transfer substitution reaction between alkyl halides (RX) and Lewis bases (HR'B). This reaction intermediate was observed for the 1-chloroadamantane and tert-butylamine system in a hydrocarbon glass at 25
Xue Li et al.
Journal of pharmaceutical sciences, 106(1), 395-401 (2016-11-07)
Supramolecular cyclodextrin-based nanoparticles (CD-NPs) mediated by host-guest interactions have gained increased popularity because of their "green" and simple preparation procedure, as well as their versatility in terms of inclusion of active molecules. Herein, we showed that original CD-NPs of around
Aerosol fluorination of 1-chloroadamantane, 2-chloroadamantane, and methyl 1-adamantylacetate: a novel synthetic approach to 1-and 2-substituted hydryl-, methyl-, and (difluoromethyl-F-adamantanes.
Adcock JL, et al.
The Journal of Organic Chemistry, 57(17), 4749-4752 (1992)
Elena L Gromnitskaya et al.
Physical chemistry chemical physics : PCCP, 23(3), 2349-2354 (2021-01-16)
We present a comparative ultrasonic study of the elastic properties of adamantane and 1-chloroadamantane at high pressure (up to 1.4 GPa) and different temperatures (77-293 K) and at order-disorder transitions. The ultrasonic method provides complementary pictures of the order-disorder transitions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service