Skip to Content
Merck
1-Chloro-2-methylpropane 98%

1-Chloro-2-methylpropane

1-Chloro-2-methylpropane 98%
Synonym(s):
Isobutyl chloride, 1-Chloro-2-methylpropane
Linear Formula:
(CH3)2CHCH2Cl
CAS Number:
Molecular Weight:
92.57

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing