Skip to Content
Merck
All Photos(2)

Key Documents

161667

Sigma-Aldrich

10-Undecenoyl chloride

97%

Synonym(s):

ψ-Undecenoyl chloride, ψ-Undecylenic acid chloride, 10-Hendecenoyl chloride, Undec-10-enoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)8COCl
CAS Number:
Molecular Weight:
202.72
Beilstein:
1635112
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

120-122 °C/10 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

ClC(=O)CCCCCCCCC=C

InChI

1S/C11H19ClO/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2H,1,3-10H2

InChI key

MZFGYVZYLMNXGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

10-Undecenoyl chloride was used as acylating reagent in the synthesis of cellulose ω-carboxyalkanoates and poly(ethylene glycol)–lipid amphiphiles. It was used in synthesis and modification of hyperbranched poly(glycidol).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huan Liu et al.
Small (Weinheim an der Bergstrasse, Germany), 16(44), e2004922-e2004922 (2020-10-09)
For the first time Janus-like films of surface-acylated cellulose nanowhiskers (CNWs) with or without graphene oxide (GO) via one-step evaporation-driven self-assembly process are reported, which have reconstructible time-dependent micro-/nanostructures and asymmetric wettability. The heterogeneous aggregation of CNWs on rough Teflon
Enhancement of the impact strength of cationically cured cycloaliphatic diepoxide by adding hyperbranched poly (glycidol) partially modified with 10-undecenoyl chains.
Flores M, et al.
Eur. Polymer J., 49(6), 1610-1620 (2013)
Xiangtao Meng et al.
Biomacromolecules, 15(1), 177-187 (2013-12-18)
Cross-metathesis has been shown for the first time to be a useful method for the synthesis of polysaccharide derivatives, focusing herein on preparation of cellulose ω-carboxyalkanoates. Commercially available cellulose esters were first acylated with 10-undecenoyl chloride, providing esters with olefin-terminated
Core crosslinkable polymeric micelles from PEG-lipid amphiphiles as drug carriers.
Tian L, et al.
Journal of Materials Chemistry, 14(14), 2317-2324 (2004)
Kamila Szwed et al.
Journal of separation science, 39(6), 1110-1117 (2016-03-31)
Cyclodextrins and their derivatives are one of the most common and successful chiral selectors. However, there have been few publications about the use of cyclodextrin-modified monoliths. In this study, organic hybrid monoliths were prepared by the immobilization of derivatized β-cyclodextrin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service