Skip to Content
Merck
All Photos(1)

Key Documents

467642

Sigma-Aldrich

11-Bromo-1-undecene

95%

Synonym(s):

1-Bromo-10-undecene, 10-Undecenyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)9CHCH2
CAS Number:
Molecular Weight:
233.19
Beilstein:
1753231
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.468 (lit.)

bp

149-150 °C/35 mmHg (lit.)

density

1.063 g/mL at 25 °C (lit.)

functional group

alkyl halide
allyl
bromo

SMILES string

BrCCCCCCCCCC=C

InChI

1S/C11H21Br/c1-2-3-4-5-6-7-8-9-10-11-12/h2H,1,3-11H2

InChI key

YPLVPFUSXYSHJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

11-Bromo-1-undecene is a halogenated hydrocarbon. It can be synthesized by employing alkenyl esters or dibromides as starting materials.

Application

11-Bromo-1-undecene to synthesize brominated polyethylene (PE) via metallocene-catalyzed copolymerization with ethylene. It is also used to prepare poly(olefin)-based anion exchange membranes (AEMs) for practical alkaline fuel cell applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 45, 931-932 (2004)
The facile preparation of alkenyl metathesis synthons.
Baughman TW, et al.
Tetrahedron, 60(48), 10943-10948 (2004)
Covalent and Non-covalent Attachment and Patterning of Polypyrrole at Silicon Surfaces.
Pike AR, et al.
Advanced Materials, 15(3), 254-257 (2003)
Synthesis of epoxy-terminated dialkyl disulfides for polymerizable self-assembled monolayers.
Yeager LJ, et al.
Tetrahedron Letters, 39(46), 8409-8412 (1998)
Marrani AG, et al.
Electrochimica Acta, 53(11), 3903-3909 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service