Skip to Content
Merck
All Photos(2)

Key Documents

156914

Sigma-Aldrich

Dimethyl sulfoxide-d6

"100%", 99.96 atom % D

Synonym(s):

(Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein:
1237248
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

0.42 mmHg ( 20 °C)

Quality Level

isotopic purity

99.96 atom % D

Assay

≥99% (CP)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %

technique(s)

NMR: suitable

impurities

≤0.0250% water
water

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfoxide-d6 (DMSO-d6) is a deuterated solvent. It is 100% isotopically enriched NMR (Nuclear Magnetic Resonance) solvent. It is widely employed in high resolution NMR studies due to its high chemical and isotopic purity. On photoirradiation in the range of 193 and 222nm, it undergoes decomposition to afford CD3 radicals. Quantum yields of CD3 have been evaluated by infrared diode laser absorption spectroscopy. 100% DMSO-d6 has been used as solvent in the long-range COSY (Correlation Spectroscopy) experiment.

Application

Dimethyl sulfoxide-d6 has been used as solvent for the dissolution of glucan during its proton, 13C (Carbon-13) and 2D NMR (Two-Dimensional Nuclear Magnetic Resonance) spectral studies. Dimethyl sulfoxide-d6 may be used as a solvent to study the nuclear magnetic resonance spectra(NMR) of the corresponding 1,3- benzoxazines and 1,3-benzoxazinones, which tend to possess antimicrobial activity.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis and antimicrobial activity of new benzofuranyl-1, 3-benzoxazines and 1, 3-benzoxazin-2-ones"
Ujjinamatada.KR, et al.
Journal of Heterocyclic Chemistry, 43(02), 437-441 (2006)
Douglas W Lowman et al.
PloS one, 6(11), e27614-e27614 (2011-11-19)
β-Glucan is a (1→3)-β-linked glucose polymer with (1→6)-β-linked side chains and a major component of fungal cell walls. β-Glucans provide structural integrity to the fungal cell wall. The nature of the (1-6)-β-linked side chain structure of fungal (1→3,1→6)-β-D-glucans has been
Miles Montgomery et al.
ACS biomaterials science & engineering, 4(11), 3691-3703 (2018-11-12)
Using the methods described herein, we have demonstrated how scaffolds can be designed for a number of applications including tissue engineering, biomedical devices and injectable tissues. Details on the methods of polymerization and physical and chemical characterization of poly(octamethylene maleate
Derek K O'Flaherty et al.
Bio-protocol, 10(17), e3734-e3734 (2021-03-05)
The RNA world hypothesis describes a scenario where early life forms relied on RNA to govern both inheritance and catalyze useful chemical reactions. Prior to the emergence of enzymes capable of replicating the RNA genome, a nonenzymatic replication process would
Lulu Wang et al.
Bioscience reports, 41(5) (2021-04-13)
Interleukin-8 (IL-8) promotes cell homing and angiogenesis, but its effects on activating human bone marrow mesenchymal stem cells (BMSCs) and promoting angiogenesis are unclear. We used bioinformatics to predict these processes. In vitro, BMSCs were stimulated in a high-glucose (HG)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service