Merck
Nonenzymatic RNA-templated Synthesis of N3'→P5' Phosphoramidate DNA.
Bio-protocol (2021-03-05)
Derek K O'Flaherty, Lijun Zhou, Jack W Szostak
ABSTRACT
The RNA world hypothesis describes a scenario where early life forms relied on RNA to govern both inheritance and catalyze useful chemical reactions. Prior to the emergence of enzymes capable of replicating the RNA genome, a nonenzymatic replication process would have been necessary to initiate Darwinian Evolution. However, the one-pot nonenzymatic RNA chemical copying of templates with mixed-sequences is insufficient to generate strand products long enough to encode useful function. The use of alternate (RNA-like) genetic polymers may overcome hurdles associated with RNA copying, and further our understanding of nonenzymatic copying chemistry. This protocol describes the nonenzymatic copying of RNA templates into N3'→P5' phosphoramidate DNA (3'-NP-DNA). We describe, in detail, the synthesis of 3'-amino-2',3'-dideoxyribonucleotide monomers activated with 2-aminoimidazole (3'-NH2-2AIpddN), and their use in template-directed polymerization.
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture