
Dimethyl sulfoxide-d6

Synonym(s):
(Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide
Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing