Skip to Content
Merck
All Photos(3)

Key Documents

175943

Sigma-Aldrich

Dimethyl sulfoxide-d6

99.5 atom % D

Synonym(s):

(Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein:
1237248
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

0.42 mmHg ( 20 °C)

Quality Level

isotopic purity

99.5 atom % D

Assay

99% (CP)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %

technique(s)

NMR: suitable

impurities

≤0.0250% water
water

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfoxide-d6 (DMSO-d6) is a deuterated solvent. On photoirradiation in the range of 193 and 222nm, it undergoes decomposition to afford CD3 radicals. Quantum yields of CD3 have been evaluated by infrared diode laser absorption spectroscopy. 100% DMSO-d6 has been used as solvent in the long-range COSY (Correlation Spectroscopy) experiment.

Application

DMSO-d6 along with glycerol or glycerol-d8 forms highly viscous binary solvents with high dissolution power for complex biological active compounds.
Dimethyl sulfoxide-d6 (DMSO-d6) has been employed as solvent for the dissolution of 1-allyloxy-2-hydroxy-propyl-starch (AHP-starch) to record its high resolution 1H NMR (Proton Nuclear Magnetic Resonance) spectrum.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Optimization of the synthesis of 1-allyloxy-2-hydroxy-propyl-starch through statistical experimental design.
Huijbrechts AML, et al.
Carbohydrate Polymers, 77(5), 25-31 (2009)
Highly Viscous Binary Solvents: DMSO-d6/Glycerol and DMSO-d6/Glycerol-d8 for Polar and Apolar Mixture Analysis by NMR.
Lameiras P and Nuzillard J-M.
Analytical Chemistry, 88(8), 4508-4515 (2016)
Akira Minami et al.
Scientific reports, 11(1), 3302-3302 (2021-02-10)
Reduction of elastin in the skin causes various skin diseases as well as wrinkles and sagging with aging. Sialidase is a hydrolase that cleaves a sialic acid residue from sialoglycoconjugate. Cleavage of sialic acid from microfibrils by the sialidase isozyme
M G Nair et al.
Applied and environmental microbiology, 57(2), 434-439 (1991-02-01)
Two isoflavonoids isolated from clover roots grown under phosphate stress were characterized as formononetin (7-hydroxy,4'-methoxy isoflavone) and biochanin A (5,7-dihydroxy,4'-methoxy isoflavone). At 5 ppm, these compounds stimulated hyphal growth in vitro and root colonization of an undescribed vesicular-arbuscular mycorrhiza, a
Using high-performance quantitative NMR (HP-qNMR?) for certifying traceable and highly accurate purity values of organic reference materials with uncertainties< 0.1%.
Weber M, et al.
Accreditation and Quality Assurance, 18(2), 91-98 (2013)

Related Content

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service