Skip to Content
Merck
All Photos(1)

Key Documents

156884

Sigma-Aldrich

3-Nitrophthalic anhydride

98%

Synonym(s):

3-Nitrophthalic acid anhydride, 4-Nitro-2-benzofuran-1,3-dione, 4-Nitroisobenzofuran-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H3NO5
CAS Number:
Molecular Weight:
193.11
Beilstein:
179963
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

mp

163-165 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc2C(=O)OC(=O)c12

InChI

1S/C8H3NO5/c10-7-4-2-1-3-5(9(12)13)6(4)8(11)14-7/h1-3H

InChI key

ROFZMKDROVBLNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactions with aminoquinazolinones yield phthalimidoquinazolinones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Serb. Chem. Soc., 57, 629-629 (1992)
Koichiro Teshima et al.
Journal of pharmaceutical and biomedical analysis, 47(3), 560-566 (2008-03-04)
A liquid chromatography-tandem mass spectrometric (LC-MS/MS) method following chemical derivatization with 3-nitrophtalic anhydride was developed for the simultaneous determination of farnesol and geranylgeraniol in rat liver and testis. One analogue compound of the analytes, n-pentadecanol, was used as an internal
M Pawlowska et al.
Journal of chromatography. A, 666(1-2), 485-491 (1994-04-22)
New pre-column derivatizing reagents: phthalic anhydride, 3-nitrophthalic anhydride, diphenic anhydride, 1,8-naphthalic anhydride and diphenylmaleic anhydride have been developed for resolving chiral compounds having amine groups. Although all of these agents produce derivatives with high molar absorptivities, the later two also
Mauro Vieira de Almeida et al.
Chemical & pharmaceutical bulletin, 55(2), 223-226 (2007-02-03)
Fourteen thalidomide analogs bearing two phthalimido units were prepared in high yields (83-94%) by condensation of different diamines with phthalic or 3-nitrophthalic anhydride. An in vitro investigation of the compounds as inhibitors of the TNF-alpha production was performed. The inhibition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service