Skip to Content
Merck
All Photos(1)

Key Documents

215155

Sigma-Aldrich

Hydrazine

anhydrous, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2
CAS Number:
Molecular Weight:
32.05
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

vapor density

1 (37 °C, vs air)

vapor pressure

10 mmHg ( 30.7 °C)

Assay

98%

form

liquid

expl. lim.

99.99 %

refractive index

n20/D 1.47 (lit.)

bp

113.5 °C (lit.)

mp

1.4 °C (lit.)

density

1.021 g/mL at 25 °C (lit.)

SMILES string

NN

InChI

1S/H4N2/c1-2/h1-2H2

InChI key

OAKJQQAXSVQMHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrazine is the simplest diamine mainly used as a reducing agent.
Sensitive determination of hydrazine with detection limit of 10.0nM has been achieved by employing an electrochemical sensor. Hydrazine undergoes dehydrogenation in the presence of bimetallic NiRh nanoparticles (NPs) supported on nitrogen-doped porous carbon (NPC) derived from metal-organic frameworks (ZIF-8) to afford hydrogen.

Application

Hydrazine has been used as one of the reaction components in the synthesis of a soluble chalcogenidometallate cluster salt, which is a precursor for the preparation of SnSe2.
Hydrazine may be used in the synthesis of:
  • pyridyl aminated O-linked sugar chains from glycoproteins
  • nanocomposite of manganese oxide embedded in graphene sheets (GS–Mn3O4)
  • formyl hydrazine by reacting with ethyl formate

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Buse Demirkan et al.
Journal of colloid and interface science, 552, 432-438 (2019-06-04)
In this study, a facile ex situ synthesis of a polyaniline-multiwalled carbon nanotube-based Pt nanocatalyst (Pt@PANI-MWCNT) with an average particle size of 3.18 ± 0.12 nm was performed successfully. The obtained Pt@PANI-MWCNT nanocatalysts were isolated from the solution medium by centrifugation and then
NiRh nanoparticles supported on nitrogen-doped porous carbon as highly efficient catalysts for dehydrogenation of hydrazine in alkaline solution.
Xia B, et al.
Nano Research, 8(11), 3472-3479 (2015)
Hadi Mahmoudi Moghaddam et al.
Environmental monitoring and assessment, 187(7), 407-407 (2015-06-06)
Hydrazine and phenol are two important environmental pollutants. In this work, an electrochemical sensor for the selective and sensitive detection of hydrazine in presence of phenol was developed by the bulk modification of carbon paste electrode (CPE) with TiO2 nanoparticles
Rapid microwave-assisted synthesis of Mn 3 O 4?graphene nanocomposite and its lithium storage properties
Li L, et al.
Journal of Materials Chemistry, 22(8), 3600-3605 (2012)
Release of O-linked sugar chains from glycoproteins with anhydrous hydrazine and pyridylamination of the sugar chains with improved reaction conditions.
Kuraya N and Hase S.
Journal of Biochemistry, 112(1), 122-126 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service