Skip to Content
Merck
All Photos(3)

Key Documents

B4600

Sigma-Aldrich

1,2,4-Benzenetricarboxylic anhydride

97%

Synonym(s):

Trimellitic anhydride, Benzene-1,2,4-tricarboxylic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4O5
CAS Number:
Molecular Weight:
192.13
Beilstein:
9394
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

6.6 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

97%

form

solid

expl. lim.

7 %

mp

163-166 °C (lit.)

SMILES string

OC(=O)c1ccc2C(=O)OC(=O)c2c1

InChI

1S/C9H4O5/c10-7(11)4-1-2-5-6(3-4)9(13)14-8(5)12/h1-3H,(H,10,11)

InChI key

SRPWOOOHEPICQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,4-Benzenetricarboxylic anhydride(TA) is a hydrophilic monomer with multipurpose usage; as a curing agent for epoxy based resins and as a plasticizer for polyvinyl chloride(PVC).

Application

TA can chemically modify sugarcane bagasse, which can be used in the absorption of auramine and safranin dyes from aqueous solutions. It can be blended with epoxy propane in the synthesis of polyurethane acrylates, which can be used as oligomers for ultra-violet based coatings. Polyamide P84 ultra-filtration membranes can be functionalized with TA by interacting the amide linkages with the anhydrides.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

440.6 °F - closed cup

Flash Point(C)

227 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Trimellitated sugarcane bagasse: A versatile adsorbent for removal of cationic dyes from aqueous solution. Part I: Batch adsorption in a monocomponent system.
Fideles RA, et al.
Journal of Colloid and Interface Science, 515(11), 172-188 (2018)
Optimized synthesis and properties of surfactant-free water-reducible acrylate-alkyd resin emulsion.
Liang L, et al.
Progress in Organic Coatings, 77(11), 1715-1723 (2014)
Vibrational, electronic and quantum chemical studies of 1, 2, 4-benzenetricarboxylic-1, 2-anhydride.
Arjunan V, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 110(11), 141-150 (2013)
Hybrid polymer/MOF membranes for Organic Solvent Nanofiltration (OSN): Chemical modification and the quest for perfection.
Campbell J.
Journal of Membrane Science , 503(2), 166-176 (2016)
Synthesis and characterization of hyperbranched polyurethane acrylates used as UV curable oligomers for coatings.
Xu G and Shi W
Progress in Organic Coatings, 52(2), 110-117 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service