Skip to Content
Merck
All Photos(1)

Key Documents

137820

Sigma-Aldrich

3-Nitrophthalic acid

99%

Synonym(s):

3-Nitro-1,2-benzenedicarboxylic acid, m-Nitrophthalic acid, o-Nitrophthalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3-1,2-(CO2H)2
CAS Number:
Molecular Weight:
211.13
Beilstein:
2054269
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

210 °C (dec.) (lit.)

solubility

water: soluble 5%, clear to slightly hazy, colorless to faint yellow or tan

functional group

carboxylic acid
nitro

SMILES string

OC(=O)c1cccc(c1C(O)=O)[N+]([O-])=O

InChI

1S/C8H5NO6/c10-7(11)4-2-1-3-5(9(14)15)6(4)8(12)13/h1-3H,(H,10,11)(H,12,13)

InChI key

KFIRODWJCYBBHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Nitrophthalic acid is a degradation product of isoxaben herbicide. It acts as ligand and forms rare-earth complexes.

Application

3-Nitrophthalic acid was used as starting reagent in the synthesis of modified 2-iodoxybenzoic acid derivatives.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fan Zhang et al.
Guang pu xue yu guang pu fen xi = Guang pu, 29(10), 2773-2776 (2009-12-30)
A series of rare-earth compound RE2L2 (HL)2 (H2O)6 x 2H2O (RE=La, Nd, Eu, Tb, Er, Y) containing 3-nitrophthalic acid (H2L) ligand were synthesized from ethanol-water solution, and characterized and investigated by the element analysis, infrared and ultraviolet absorption spectra and
Clyatt E Drakeford et al.
Chemosphere, 50(9), 1243-1247 (2003-01-28)
Commercial production of ornamental plants is an important industry in the United States and involves a complex technology that includes the use of herbicides. Isoxaben[N-[3-(1-ethyl-1-methylpropyl)-5-isoxazolyl]-2,6-dimethoxybenzamide] is a pre-emergence herbicide used for controlling weeds in many areas including containerized ornamental plants.
Hypervalent iodine reagents for the oxidation of alcohols and their application to complex molecule synthesis.
Tohma H and Kita Y.
Advanced Synthesis & Catalysis, 346(2-3), 111-124 (2004)
En-Cui Yang et al.
Dalton transactions (Cambridge, England : 2003), 44(7), 3190-3199 (2015-01-13)
Four new 4-amino-1,2,4-triazole (atr)-based coordination polymers, {[Cu2(atr)(H2O)(μ-OH)2(pa)]·H2O}n (), {[Cu3(atr)2(H2O)2(μ-OH)2(npa)2]·2H2O}n (), {[Cu3(atr)5(dca)(μ-OH)(ClO4)2](ClO4)2}n () and {[Cu3(atr)2(H2O)(μ3-OH)(μ-OH)2(spa)]·1.5H2O}n () (pa(2-) = phthalate, npa(2-) = 3-nitrophthalate, dca(-) = dicyanamide and spa(3-) = 4-sulfophthalate), were successfully obtained by varying the carboxylate- and cyanide-modified magnetic bridges. Structural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service