Skip to Content
Merck
All Photos(1)

Key Documents

14050

Sigma-Aldrich

Berberine chloride hydrate

technical, ≥90% (AT)

Synonym(s):

Natural Yellow 18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18ClNO4 · xH2O
CAS Number:
Molecular Weight:
371.81 (anhydrous basis)
Colour Index Number:
75160
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (AT)

form

powder

mp

204-206 °C (dec.)

SMILES string

[Cl-].COc1ccc2cc3-c4cc5OCOc5cc4CC[n+]3cc2c1OC

InChI

1S/C20H18NO4.ClH/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;/h3-4,7-10H,5-6,11H2,1-2H3;1H/q+1;/p-1

InChI key

VKJGBAJNNALVAV-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sayan Biswas et al.
Pharmaceutical development and technology, 26(1), 69-80 (2020-10-17)
Piper species is one of the most widely consumed spices for culinary purposes. Piperine (PIP) present in Piper species has a wide range of therapeutic activity including hepatoprotection. However, the major biological limitation of PIP is its low bioavailability after
Lamyae El Khalki et al.
Molecules (Basel, Switzerland), 25(3) (2020-01-30)
Triple negative breast cancer (TNBC) is an aggressive breast cancer subtype. Non-available targeted therapy for TNBC represents its biggest treatment challenge. Thus, finding new promising effective drugs is urgently needed. In the present study, we investigated how berberine, a natural
Li Jia et al.
Mediators of inflammation, 2020, 9419085-9419085 (2020-10-17)
Berberine (BER), a natural isoquinoline alkaloid, has been demonstrated to have appreciable anticolitis effects. Nevertheless, the protective mechanism of BER in ulcerative colitis (UC) is barely understood. The present study was aimed at exploring the therapeutic efficacy of BER on
Cemile Selin Aksoy et al.
Microbial pathogenesis, 149, 104542-104542 (2020-10-04)
A plethora of natural products emerges as attractive molecules in the struggle against antibiotic resistance. These molecules impose their bioactivities not only alone but also in combinations as well, which further enhances their effects. Berberine is a well-known isoquinoline alkaloid
Victor P Bagla et al.
BMC complementary and alternative medicine, 14, 383-383 (2014-10-09)
Different parts of Podocarpus henkelii have been used in many cultures around the world to treat ailments such as cholera, stomach diseases, rheumatism, cancer, canine distemper in dogs and gall sickness in cattle. The aim of this study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service