Skip to Content
Merck
All Photos(1)

Key Documents

SML0539

Sigma-Aldrich

T863

≥98% (HPLC)

Synonym(s):

2-((1,4-trans)-4-(4-(4-Amino-7,7-dimethyl-7H-pyrimido[4,5-b][1,4]oxazin-6-yl)- phenyl)cyclohexyl)acetic acid, T-863, trans-4-[4-(4-Amino-7,7-dimethyl-7H-pyrimido[4,5-b][1,4]oxazin-6-yl)phenyl]-cyclohexaneacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N4O3
CAS Number:
Molecular Weight:
394.47
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

CC1(C)Oc2ncnc(N)c2N=C1c3ccc(cc3)[C@@H]4CC[C@H](CC4)CC(O)=O

InChI

1S/C22H26N4O3/c1-22(2)19(26-18-20(23)24-12-25-21(18)29-22)16-9-7-15(8-10-16)14-5-3-13(4-6-14)11-17(27)28/h7-10,12-14H,3-6,11H2,1-2H3,(H,27,28)(H2,23,24,25)/t13-,14-

InChI key

FUIYMYNYUHVDPT-HDJSIYSDSA-N

Application

T863 has been used as a diglyceride acyltransferase (DGAT1) inhibitor:
  • to provide the opportunity to precisely control the DGAT inhibition
  • to treat mouse embryonic fibroblasts (MEFs) to analyse lipid droplet biogenesis
  • to specifically block neutral lipid synthesis and lipid droplet formation in HT-1080 cells
  • to block lipid droplet (LD) formation

Biochem/physiol Actions

T863 is an orally active, selective and potent DGAT1 (Acyl-CoA:diacylglycerol acyltransferase 1) inhibitor that interacts with the acyl-CoA binding site of DGAT1, and inhibits triacylglycerol synthesis in cells. T863 causes weight loss, reduction in serum and liver triglycerides, and improved insulin sensitivity in obese mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DGAT1-dependent lipid droplet biogenesis protects mitochondrial function during starvation-induced autophagy
Nguyen TB, et al.
Developmental Cell, 42(1), 9-21 (2017)
Lipid droplet formation in Mycobacterium tuberculosis infected macrophages requires IFN gamma/HIF-1alpha signaling and supports host defense
Knight M, et al.
PLoS Pathogens, 14(1), e1006874-e1006874 (2018)
Angela Castoldi et al.
Nature communications, 11(1), 4107-4107 (2020-08-17)
Foamy macrophages, which have prominent lipid droplets (LDs), are found in a variety of disease states. Toll-like receptor agonists drive triacylglycerol (TG)-rich LD development in macrophages. Here we explore the basis and significance of this process. Our findings indicate that
Exogenous monounsaturated fatty acids promote a ferroptosis-resistant cell state
Magtanong L, et al.
Cell Chemical Biology, 26(3), 420-432 (2019)
Jesus A Silvas et al.
bioRxiv : the preprint server for biology (2020-08-04)
Therapeutics targeting replication of SARS coronavirus 2 (SARS-CoV-2) are urgently needed. Coronaviruses rely on host membranes for entry, establishment of replication centers and egress. Compounds targeting cellular membrane biology and lipid biosynthetic pathways have previously shown promise as antivirals and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service