Skip to Content
Merck
All Photos(1)

Key Documents

S5890

Sigma-Aldrich

Sanguinarine chloride hydrate

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14ClNO4 · xH2O
CAS Number:
Molecular Weight:
367.78 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

solubility

H2O: slightly soluble <0.3 mg/mL
methanol: 3.8 mg/mL

SMILES string

[Cl-].C[n+]1cc2c3OCOc3ccc2c4ccc5cc6OCOc6cc5c14

InChI

1S/C20H14NO4.ClH/c1-21-8-15-12(4-5-16-20(15)25-10-22-16)13-3-2-11-6-17-18(24-9-23-17)7-14(11)19(13)21;/h2-8H,9-10H2,1H3;1H/q+1;/p-1

InChI key

GIZKAXHWLRYMLE-UHFFFAOYSA-M

Related Categories

Application

Sanguinarine chloride hydrate was tested for anti-schistosomal activities against Schistosoma mansoni.9 It was tested for anti-lipase activity against Candida rugosa lipase.10
Sanguinarine chloride hydrate yields clear, orange solution in methanol at 20 mg/ml.

Biochem/physiol Actions

A natural product with antimicrobial, anti-inflammatory, and anti-oxidant properties.
A natural product with antimicrobial, anti-inflammatory, and anti-oxidant properties. Has antiproliferative, pro-apoptosis effects in some cancer cell lines.
Sanguinarine is a benzophenanthridine alkaloid isolated from plants belonging to the family Papaveracea. It exhibits anti-bacterial, anti-fungal, anti-inflammatory and anti-cancer properties. It induces cell cycle arrest and sensitizes cancer cells to apoptosis by activating TNF-related apoptosis inducing ligand.6,7 It inhibits STAT3, MMP-2, MMP-9, interacts with glutathione, induces generation of ROS, disrupts the microtubule assembly and causes DNA damage resulting the death of the cancer cells.6,7,8

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Jian Lu et al.
Evidence-based complementary and alternative medicine : eCAM, 2012, 485042-485042 (2012-09-19)
Alkaloids are important chemical compounds that serve as a rich reservoir for drug discovery. Several alkaloids isolated from natural herbs exhibit antiproliferation and antimetastasis effects on various types of cancers both in vitro and in vivo. Alkaloids, such as camptothecin
Hieu Huu Le et al.
Antioxidants (Basel, Switzerland), 9(10) (2020-10-25)
Heat stress (HS) compromises productivity of pork production, in part as a result of increased oxidative stress and inflammatory responses, particularly within the gastrointestinal tract. This study aimed to investigate whether plant-derived betaine and isoquinoline alkaloids could ameliorate HS in
Matthew Mendoza Barker et al.
Journal of fungi (Basel, Switzerland), 10(2) (2024-02-23)
Coccidioidomycosis (CM) can manifest as respiratory and disseminated diseases that are caused by dimorphic fungal pathogens, such as Coccidioides species. The inhaled arthroconidia generated during the saprobic growth phase convert into multinucleated spherules in the lungs to complete the parasitic
Christian Vlachojannis et al.
Phytotherapy research : PTR, 26(10), 1423-1426 (2012-02-10)
The rhizome of Sanguinaria canadensis (SC, bloodroot) contains an active principle with antimicrobial, antiinflammatory, antioxidative and immunomodulatory effects. For this reason SC extract has been added to toothpastes and mouthwashes in various concentrations. When tested separately, neither the toothpastes nor
Min Ho Han et al.
Toxicology letters, 220(2), 157-166 (2013-05-11)
We examined the effects of sanguinarine, a benzophenanthridine alkaloid, on reactive oxygen species (ROS) production and the association of these effects with apoptotic cell death in a human colorectal cancer HCT-116 cell line. Sanguinarine generated ROS, which was followed by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service