Skip to Content
Merck
All Photos(5)

Key Documents

B3251

Sigma-Aldrich

Berberine chloride form

Synonym(s):

Benzodioxide, Berberine HCl, Natural Yellow 18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18ClNO4
CAS Number:
Molecular Weight:
371.81
Colour Index Number:
75160
Beilstein:
3836585
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

form

powder

Quality Level

solubility

methanol: soluble

SMILES string

COC1=C(C=[N+](CCC2=C3C=C4C(OCO4)=C2)C3=C5)C5=CC=C1OC.[Cl-]

InChI

1S/C20H18NO4.ClH/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;/h3-4,7-10H,5-6,11H2,1-2H3;1H/q+1;/p-1

InChI key

VKJGBAJNNALVAV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Berberine Chloride form or Natural Yellow 18, is also known as 5,6-dihydro-9,10-dimethoxybenzo(g)-1,3-benzodioxolo (5,6-a) quinolizinium is a benzyl tetra isoquinoline plant alkaloid. Berberine contains a quaternary base and is commercially available in berberine chloride form.

Application

Berberine Chloride is a fluorescent stain used to stain heparin in mast cells. Berberine upregulates the expression of Pgp in hepatoma cells.
Fluorescent stain for heparin in mast cells

Biochem/physiol Actions

An alkaloid with weak antibiotic properties. Substrate for MDR efflux pumps. Antimicrobial activities of berberine is potentiated by the MDR inhibitor 5´-methoxyhydnocarpin (5´-MHC). Berberine upregulates the expression of Pgp in hepatoma cells. Treatment with berberine potentially results in the reduced accumulation of chemotherapeutic drugs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-Hong Li et al.
Journal of medicinal chemistry, 52(2), 492-501 (2008-12-19)
Twenty-nine derivatives of berberine (1) or pseudoberberine (2) were designed, semisynthesized, and evaluated for their up-regulatory activity on the low-density-lipoprotein receptor (LDLR) expression. SAR analysis revealed that (i) the methylenedioxy group at the 2- and 3-position is an essential element
Xiaoling Zhang et al.
Cancer research, 70(23), 9895-9904 (2010-10-12)
Berberine, a natural product derived from a plant used in Chinese herbal medicine, is reported to exhibit anticancer effects; however, its mechanism of action is not clearly defined. Herein, we demonstrate that berberine induces apoptosis in acute lymphoblastic leukemia (ALL)
Siritron Samosorn et al.
Bioorganic & medicinal chemistry, 17(11), 3866-3872 (2009-05-08)
Conjugation of the NorA substrate berberine and the NorA inhibitor 5-nitro-2-phenyl-1H-indole via a methylene ether linking group gave the 13-substituted berberine-NorA inhibitor hybrid, 3. A series of simpler arylmethyl ether hybrid structures were also synthesized. The hybrid 3 showed excellent
Inhibitors of prolyl oligopeptidases for the therapy of human diseases: defining diseases and inhibitors.
Janice Lawandi et al.
Journal of medicinal chemistry, 53(9), 3423-3438 (2010-01-12)
Yan Ma et al.
Bioorganic & medicinal chemistry, 16(16), 7582-7591 (2008-08-05)
A series of 9-N-substituted berberine derivatives (2a-j) were synthesized and evaluated as a new class of G-quadruplex binding ligands. G-quadruplex of DNA had been proven to be the transcription controller of human c-myc gene. The interaction of 9-N-substituted berberine derivatives

Articles

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service