Skip to Content
Merck
All Photos(3)

Key Documents

130109

Sigma-Aldrich

2-Naphthol

98%

Synonym(s):

β-Naphthol, 2-Hydroxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OH
CAS Number:
Molecular Weight:
144.17
Beilstein:
742134
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.97 (vs air)

Quality Level

vapor pressure

10 mmHg ( 145.5 °C)

Assay

98%

form

powder or flakes

bp

285-286 °C (lit.)

mp

120-122 °C (lit.)

SMILES string

Oc1ccc2ccccc2c1

InChI

1S/C10H8O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H

InChI key

JWAZRIHNYRIHIV-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Co2O3-TiO2 exhibited high visible-light activities for the degradation of 2-Naphthol (2-Hydroxynaphthalene).

Application

2-Naphthol (2-Hydroxynaphthalene) was used to study the kinetics of the recombinant Coprinus cinereus peroxidase-catalyzed 2-naphthol oxidation.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

307.4 °F - closed cup

Flash Point(C)

153 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivanec-Goranina Rūta et al.
Journal of environmental sciences (China), 25(7), 1431-1440 (2013-11-14)
The kinetics of the recombinant Coprinus cinereus peroxidase-catalyzed 2-naphthol oxidation was investigated in the presence of rhamnolipid biosurfactant JBR425 and synthetic surfactant Surfynol465 at pH 5.5 and 250C, with concentrations of (bio)surfactants both less than critical micelle concentrations (CMC) and
Qiliang Jin et al.
Physical chemistry chemical physics : PCCP, 15(46), 20313-20319 (2013-10-31)
This study first presents a "TiO2-based eco-catalyst" working in the dark and under visible-light irradiation for the degradation of environmental organic pollutants. Molecular scale cobalt(III) oxide clusters are formed on the surface of highly active anatase TiO2 nanoparticles (Co2O3-TiO2) by
Glenn Talaska et al.
Toxicology letters, 213(1), 45-48 (2011-05-26)
Urinary bladder cancer is a historical disease of rubber workers often been associated with exposure to aromatic amines such as 2-naphthylamine. While exposure to these compounds has decreased markedly over time, the bladder cancer risk has not decreased in direct
Erin E Podlesny et al.
Organic letters, 14(6), 1408-1411 (2012-03-01)
The first enantioselective total synthesis of the bisanthraquinone (S)-bisoranjidiol and an unnatural regioisomer has been accomplished. Key features of the synthesis include the asymmetric oxidative biaryl coupling of a hindered 8-substituted 2-naphthol, selective para-quinone formation, and regioselective tandem Diels-Alder/aromatization reactions.
Supriti Sen et al.
The Analyst, 137(17), 3975-3981 (2012-07-13)
An efficient water soluble fluorescent Al(3+) receptor, 1-[[(2-furanylmethyl)imino]methyl]-2-naphthol (1-H) was synthesized and characterized by physico-chemical and spectroscopic tools along with single crystal X-ray crystallography. High selectivity and affinity of 1-H towards Al(3+) in HEPES buffer (DMSO/water: 1/100) of pH 7.4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service