Skip to Content
Merck
All Photos(4)

Key Documents

D131350

Sigma-Aldrich

1,4-Dimethoxybenzene

ReagentPlus®, 99%

Synonym(s):

p-Methoxyanisole, Dimethylhydroquinone, Hydroquinone dimethyl ether, NSC 7483

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
Beilstein:
774605
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.8 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

crystals

autoignition temp.

1463 °F

bp

213 °C (lit.)

mp

54-56 °C (lit.)

density

1.053 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(OC)cc1

InChI

1S/C8H10O2/c1-9-7-3-5-8(10-2)6-4-7/h3-6H,1-2H3

InChI key

OHBQPCCCRFSCAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Masuda et al.
Bioscience, biotechnology, and biochemistry, 60(5), 806-810 (1996-05-01)
Bovine serum albumin (BSA) added to the reaction medium for the oxidative demethylation of 4-methoxyanisole and its "di-CD3" isotopomer ([d6]methoxyanisole), when catalyzed by liver microsomes from untreated rats, decreased the Km values and increased the V(max)/Km (= V/K) values. The
Tiantian Li et al.
The journal of physical chemistry. A, 115(19), 4988-4994 (2011-04-27)
The effect of substituents on the oxidation potential for the one-electron reaction of 1,4-dimethoxybenzene was understood with a theoretical calculation based on density functional theory (DFT) at the level of B3LYP/6-311+G(d). It is found that the oxidation potential for the
D W Kim et al.
Organic letters, 3(3), 445-447 (2001-06-29)
[figure: see text] Fused 1,4-dimethoxybenzenes could be oxidized to benzoquinones by either direct oxidation or demethylation-oxidation. The oxidative demethylation of 5,8-dimethoxy-2-methylquinoline using 1.1 equiv of NBS in aqueous THF and a catalytic amount of H2SO4 at 20 degrees C for
H J Altmann et al.
Archives of toxicology. Supplement. = Archiv fur Toxikologie. Supplement, 8, 114-116 (1985-01-01)
Butylated hydroxyanisole (BHA) is widely used as an antioxidant in foodstuffs, in materials which come into contact with food and also in cosmetic products. The safety of BHA was questioned, however, when it was reported that in a recent Japanese
Decrease in glucose oxidation in isolated brown fat cells from rats due to tropolone and dimethoxybenzene.
J W Rosenthal
General pharmacology, 12(1), 47-50 (1981-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service