Skip to Content
Merck
All Photos(1)

Key Documents

185434

Sigma-Aldrich

1,1,2,2-Tetrachloroethane

reagent grade, ≥98%

Synonym(s):

Acetylene tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CHCl2CHCl2
CAS Number:
Molecular Weight:
167.85
Beilstein:
969206
EC Number:
MDL number:
UNSPSC Code:
12352101
eCl@ss:
39050126
PubChem Substance ID:
NACRES:
NA.04
grade:
reagent grade
Assay:
≥98%
bp:
147 °C (lit.)
vapor pressure:
8 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor density

5.8 (vs air)

vapor pressure

8 mmHg ( 20 °C)

Assay

≥98%

form

liquid

refractive index

n20/D 1.494 (lit.)

bp

147 °C (lit.)

mp

−43 °C (lit.)

density

1.586 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)C(Cl)Cl

InChI

1S/C2H2Cl4/c3-1(4)2(5)6/h1-2H

InChI key

QPFMBZIOSGYJDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1,2,2-Tetrachloroethane (TeCA) is a chlorine containing hydrocarbon solvent useful for dissolution of cellulose acetate, fat, waxes, greases, rubber and sulfur. It is widely used in the manufacturing of polymers.[1] Among the chlorinated hydrocarbons, it has the highest solvent power.[2]

Application

1,1,2,2-Tetrachloroethane may be employed as a solvent additive for 2,2′,7,7′-tetrakis-(N,N-di-p-methoxyphenyl-amine)9,9′spiro-bifluorene (Spiro-OMeTAD). Spiro-OMe TAD is a triarylamine-based organic p-type organic hole transport material (HTM) employed in solid-state dye-sensitized solar cells.[3]

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia C Hestenes et al.
Frontiers in chemistry, 8, 681-681 (2020-08-28)
Performance decline in Li-excess cathodes is generally attributed to structural degradation at the electrode-electrolyte interphase, including transition metal migration into the lithium layer and oxygen evolution into the electrolyte. Reactions between these new surface structures and/or reactive oxygen species in
Jian-Hong Tang et al.
Nature communications, 10(1), 4599-4599 (2019-10-12)
Host-guest interactions are of central importance in many biological and chemical processes. However, the investigation of the formation and decomplexation of host-guest systems at the single-molecule level has been a challenging task. Here we show that the single-molecule conductance of
Haoyu Li et al.
Journal of the American Chemical Society, 143(1), 481-487 (2020-12-29)
Polysulfide anions are endowed with unique redox properties, attracting considerable attentions for their applications in alkali metals-sulfur batteries. However, the employment of these anionic species in redox catalysis for small molecule synthesis remains underdeveloped due to their moderate-poor electrochemical potential
Jingjing Li et al.
Cold Spring Harbor protocols, 2019(4) (2018-06-14)
The African clawed frog Xenopus has remarkable capacities to heal wounds rapidly and to regenerate complex tissues. Because of its experimental tractability, studies using Xenopus oocytes, embryos, and larvae have contributed extensively to our understanding of the molecular and cellular
Transformations of 1, 1, 2, 2-tetrachloroethane under methanogenic conditions.
Chen C, et al.
Environmental Science & Technology, 30(2), 542-547 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service