Skip to Content
Merck
All Photos(1)

Key Documents

126136

Sigma-Aldrich

Diethyl methylmalonate

99%

Synonym(s):

Methylmalonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(COOC2H5)2
CAS Number:
Molecular Weight:
174.19
Beilstein:
637028
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.413 (lit.)

bp

198-199 °C (lit.)
78-80 °C/10 mmHg

density

1.022 g/mL at 20 °C (lit.)

SMILES string

CCOC(=O)C(C)C(=O)OCC

InChI

1S/C8H14O4/c1-4-11-7(9)6(3)8(10)12-5-2/h6H,4-5H2,1-3H3

InChI key

UPQZOUHVTJNGFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl methylmalonate reacts with 2-cyclohexenone under abnormal Michael conditions form 2-(3-oxocyclohexyl)-2-cyclohexenone. It causes the alkylation of poly(chloromethylstyrene) during pahse transfer catalysis.

Application

Diethyl methylmalonate was used to investigate the effect of direct supplementation of the medium with di-ester of methylmalonate on erythromycin fermentation in Saccharopolyspora erythraea mutB strain.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alkylation reaction of poly (chloromethylstyrene) with malononitrile and diethyl methylmalonate using phase transfer catalysts.
Nishikubo T, et al.
Makromol. Chem., 2(6-7), 387-392 (1981)
J Mark Weber et al.
Applied microbiology and biotechnology, 93(4), 1575-1583 (2011-11-04)
The Saccharopolyspora erythraea mutB knockout strain, FL2281, having a block in the methylmalonyl-CoA mutase reaction, was found to carry a diethyl methylmalonate-responsive (Dmr) phenotype in an oil-based fermentation medium. The Dmr phenotype confers the ability to increase erythromycin A (erythromycin)
Abnormal Michael reaction. Reaction between 2-cyclohexenone and diethyl methylmalonate.
Risinger GE and Haag WG.
The Journal of Organic Chemistry, 38(20), 3546-3647 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service