Skip to Content
Merck
All Photos(1)

Key Documents

W237507

Sigma-Aldrich

Diethyl malonate

≥98%, FG

Synonym(s):

Malonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOC2H5)2
CAS Number:
Molecular Weight:
160.17
FEMA Number:
2375
Beilstein:
774687
EC Number:
Council of Europe no.:
2106
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.490
NACRES:
NA.21
Organoleptic:
apple; green; fruity; sweet
grade:
FG
Halal
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

5.52 (vs air)

vapor pressure

1 mmHg ( 40 °C)

Assay

≥98%

refractive index

n20/D 1.413 (lit.)

bp

199 °C (lit.)

mp

−51-−50 °C (lit.)

density

1.055 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; green; fruity; sweet

SMILES string

CCOC(=O)CC(=O)OCC

InChI

1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3

InChI key

IYXGSMUGOJNHAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Diethyl malonate is an active methylene compound that can be used as a flavoring agent in the food industry.

Application


  • Synthesis and biological activities of novel structural analogues of 2-arachidonoylglycerol, an endogenous cannabinoid receptor ligand.: This study by Suhara et al. focuses on the synthesis of novel structural analogues of 2-arachidonoylglycerol, an endogenous ligand for cannabinoid receptors. The research highlights the utilization of diethyl malonate in the synthesis process, emphasizing its importance in creating biologically active compounds with potential therapeutic applications in modulating cannabinoid receptor activity (Suhara et al., 2001).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encyclopedia of Food and Color Additives, 1, 818-818 (1997)
Encyclopedia of Food and Color Additives, 1, 818-818 (1997)
Alessandro Marrone et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(43), 11537-11550 (2009-09-25)
The condensation of dialkyl beta-diesters with various aldehydes promoted by TiCl4 has been studied by DFT approaches and experimental methods, including NMR, IR and UV/Vis spectroscopy. Various possible reaction pathways have been investigated and their energy profiles evaluated to find
Renhua Fan et al.
The Journal of organic chemistry, 72(15), 5905-5907 (2007-06-26)
A highly efficient, one-flask tandem Knoevenagel-Michael addition reaction of sulfonimines with diethyl malonate in the presence of a catalytic amount of base affords the corresponding arylidene dimalonates in good to excellent yields.
Hua Li et al.
Nucleosides, nucleotides & nucleic acids, 29(8), 581-590 (2010-07-28)
This article describes a very simple route for synthesizing novel lipophilic phosphonate bis(t-bu-SATE) prodrugs of acyclic cyclopentenylated nucleosides such as adenine 17 and cytosine 18. The key intermediate 6 was constructed via a ring-closing metathesis of compound 5, which could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service