Skip to Content
Merck
All Photos(1)

Key Documents

M29908

Sigma-Aldrich

Methyl benzoate

99%

Synonym(s):

Methyl benzenecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH3
CAS Number:
Molecular Weight:
136.15
Beilstein:
1072099
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.68 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.516 (lit.)

bp

198-199 °C (lit.)

mp

−12 °C (lit.)

solubility

H2O: soluble 2.1 g/L at 20 °C

density

1.088 g/mL at 20 °C (lit.)

SMILES string

COC(=O)c1ccccc1

InChI

1S/C8H8O2/c1-10-8(9)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

QPJVMBTYPHYUOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl benzoate is a volatile aromatic ester compound widely used in perfumery industries. It is naturally occurring in guava, mango, and kiwifruit.

Application

Methyl benzoate (C6H5CO2CH3) can be utilized as a precursor for:
  • Selective synthesis of benzaldehyde using supported manganese oxide catalysts.;
  • Preparation of benzophenone derivatives by reacting with aryl compounds via Friedel-Crafts acylation.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - Pensky-Martens closed cup

Flash Point(C)

77 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogenation of methyl benzoate to benzaldehyde over manganese oxide catalysts prepared from Mg/Mn/Al hydrotalcite-like compounds
Chen A, et al.
Applied Catalysis A: General, 274(1-2), 101-109 (2004)
Characterization and Activity of MnO/γ-Al2O3 for Hydrogenation of Methyl Benzoate to Benzaldehyde
Xu H-L, et al.
Chin. J. Chem., 19(7), 647-651 (2001)
Direct aerobic oxidation of primary alcohols to methyl esters catalyzed by a heterogeneous gold catalyst
Nielsen IS, et al.
Catalysis Letters, 116(1-2), 35-40 (2007)
N Dudareva et al.
The Plant cell, 12(6), 949-961 (2000-06-15)
In snapdragon flowers, the volatile ester methyl benzoate is the most abundant scent compound. It is synthesized by and emitted from only the upper and lower lobes of petals, where pollinators (bumblebees) come in contact with the flower. Emission of
Trifluoromethanesulfonic acid catalyzed novel Friedel-Crafts acylation of aromatics with methyl benzoate.
Hwang JP, et al.
Tetrahedron, 56(37), 7199-7203 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service