Recommended Products
Quality Level
Assay
99%
refractive index
n20/D 1.422 (lit.)
bp
235-240 °C (lit.)
density
0.983 g/mL at 25 °C (lit.)
SMILES string
CCCCC(C(=O)OCC)C(=O)OCC
InChI
1S/C11H20O4/c1-4-7-8-9(10(12)14-5-2)11(13)15-6-3/h9H,4-8H2,1-3H3
InChI key
RPNFNBGRHCUORR-UHFFFAOYSA-N
General description
Diethyl malonate (DEM) undergoes alkylation by ethyliodide, which is used to study the phase transfer catalysis (PTC) technique.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
201.2 °F - closed cup
Flash Point(C)
94 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Inflammation, 43(4), 1323-1336 (2020-03-15)
Trained immunity has been recently identified in innate immune cells, which undergo long-term epigenetic and metabolic reprogramming after exposure to pathogens for protection from secondary infections. (1, 3)/(1, 6)-β-glucan derived from fungi can induce potent trained immunity; however, the effect
Microwave irradiation as an alternative to phase transfer catalysis in the liquid-solid phase, solvent-free C-alkylation of active methylene containing substrates.
Green Chemistry, 1073-1075 (2006)
Molecular cell, 73(2), 354-363 (2018-12-26)
Ferroptosis is a regulated necrosis process driven by iron-dependent lipid peroxidation. Although ferroptosis and cellular metabolism interplay with one another, whether mitochondria are involved in ferroptosis is under debate. Here, we demonstrate that mitochondria play a crucial role in cysteine-deprivation-induced
Redox biology, 36, 101619-101619 (2020-08-31)
Ferroptosis is a recently discovered form of programmed cell death, but its regulatory mechanisms are not fully understood. In the current study, we reported that the BRD7-P53-SLC25A28 axis played a crucial role in regulating ferroptosis in hepatic stellate cells (HSCs).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service