Merck
Mohs micrographic surgery for the treatment of melanoma.
Dermatologic clinics (2012-07-18)
Andrea M Hui, Michael Jacobson, Orit Markowitz, Norman A Brooks, Daniel M Siegel
ABSTRACT
For melanoma treatment, the primary goal is complete removal with histologically negative margins. Mohs micrographic surgery (MMS) has been extensively used and studied for the treatment of nonmelanoma skin cancer, particularly at sites where tissue conservation is vital. The use of MMS for melanoma treatment has yet to become widely accepted owing to difficulties in histologic interpretation, among other factors. MMS may offer lower recurrence rates and improved survival when compared with historical controls for standard excision. Continued advances in MMS technique and immunohistochemical staining have allowed the technique to gain further support.
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zinc chloride, JIS special grade, ≥98.0%
Sigma-Aldrich
Zinc chloride, SAJ first grade, ≥95.0%
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Zinc chloride 0.1 M solution
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%