Skip to Content
Merck
All Photos(1)

Key Documents

37-0290

Sigma-Aldrich

Zinc chloride

SAJ first grade, ≥95.0%

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

1 mmHg ( 428 °C)

Assay

≥95.0%

form

solid

reaction suitability

reagent type: catalyst
core: zinc

availability

available only in Japan

pH

5 (20 °C, 100 g/L)

mp

293 °C (lit.)

storage temp.

15-25°C

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuella P Kaster et al.
Pharmacological reports : PR, 64(3), 706-713 (2012-07-21)
In this work, the contribution of NMDA receptors to the antidepressant-like effect of adenosine in the forced swimming test (FST) was investigated. The pretreatment of mice with NMDA or D-serine was able to prevent the anti-immobility effect of either adenosine
Andrea M Hui et al.
Dermatologic clinics, 30(3), 503-515 (2012-07-18)
For melanoma treatment, the primary goal is complete removal with histologically negative margins. Mohs micrographic surgery (MMS) has been extensively used and studied for the treatment of nonmelanoma skin cancer, particularly at sites where tissue conservation is vital. The use
Michele K Sankako et al.
Pharmacological reports : PR, 64(6), 1537-1546 (2013-02-15)
The aim of this study was to evaluate the changes in testicular function of rats due to cigarette smoke exposure and the possible mechanism by which zinc protects against these alterations. Male Wistar rats (60 days old) were randomly divided
Masatoshi Ishida et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(45), 14329-14341 (2012-09-26)
The synthesis, characterization, photophysical properties, and theoretical analysis of a series of tetraaza porphyrin analogues (H-Pn: n=1-4) containing a dipyrrin subunit and an embedded 1,10-phenanthroline subunit are described. The meso-phenyl-substituted derivative (H-P1) interacts with a Mg(2+) salt (e.g., MgCl(2), MgBr(2)
Russell E Carter et al.
Journal of neurochemistry, 125(5), 673-684 (2013-03-19)
Spreading depolarization (SD) is a feed-forward wave that propagates slowly throughout brain tissue and recovery from SD involves substantial metabolic demand. Presynaptic Zn(2+) release and intracellular accumulation occurs with SD, and elevated intracellular Zn(2+) ([Zn(2+) ]i ) can impair cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service